Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01337701_m1
          See other 4930447K03RIK GE Assays ›
          Gene Symbol:
          4930447K03Rik
          Gene Name:
          RIKEN cDNA 4930447K03 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 34084155 - 34113272 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01337701_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930447K03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046184.1 4 - 5 531 67
          GenBank mRNA AK015408.1 - 4 - 5 531 67

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream tubulin,
          beta 2A class IIA
          Tubb2a M(beta)2,
          Tubb2
          -6.1kb Sense Chr. 13 - 34074278 - 34078008 on NCBI Build GRCm38
          Downstream tubulin,
          beta 2B class IIB
          Tubb2b 2410129E14Rik,
          brdp
          38.0kb Sense Chr. 13 - 34127008 - 34130354 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001780.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2255250 AK015408.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648260 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633918 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 392084 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 307565 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 68087 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 254989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 673718 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 260328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461885 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 361432 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 832247 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481672 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 661802 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 521779 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 250261 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 670801 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 83108 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111687 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145951 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145951 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07832876 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07832831 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02661632 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07832839 - -
          Whole Mouse Genome G4122A A_51_P234397 AK015408.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145951 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07832831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10518879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10774589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11043011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10664821 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936705 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11043012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10596721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10774590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10774600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10713793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10774595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10664841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10623649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10544408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10774593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10674874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10623647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11043003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10948954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11043014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28936709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10774604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10951047 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432366_PM_at - -
          Mouse Genome 430 2.0 Array 1432366_at - -
          Mouse Genome 430 2.0 Array 1432366_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300011113.mm.1 TC1300001780.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300011113.mm.1 NR_046184.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300011113.mm.1 NR_046184.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300011114.mm.1 TC1300001780.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300011114.mm.1 NR_046184.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300011114.mm.1 NR_046184.1 Refseq
          Mouse Expression Set 430 1432366_PM_at - -
          Mouse Expression Set 430 1432366_at - -
          Affymetrix HT Mouse Genome 430B Array 1432366_at - -
          Affymetrix HT Mouse Genome 430B Array 1432366_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 485617 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 106980152 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225767 - -
          Illumina Sentrix mouse-6.v1 bead array 106980152 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106980152 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225767 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0