Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200039020.mm.1 |
TC0200004555.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200039020.mm.1 |
NM_029629.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039020.mm.1 |
NM_029629.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039021.mm.1 |
TC0200004555.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200039021.mm.1 |
NM_029629.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039021.mm.1 |
NM_029629.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039025.mm.1 |
TC0200004555.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200039025.mm.1 |
NM_029629.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039025.mm.1 |
NM_029629.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039022.mm.1 |
TC0200004555.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200039022.mm.1 |
NM_029629.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039022.mm.1 |
NM_029629.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039018.mm.1 |
TC0200004555.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200039018.mm.1 |
NM_029629.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039018.mm.1 |
NM_029629.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039019.mm.1 |
TC0200004555.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200039019.mm.1 |
NM_029629.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200039019.mm.1 |
NM_029629.2 |
Refseq |