Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300011583.mm.1 |
TC1300001857.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300011583.mm.1 |
NM_001145802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011583.mm.1 |
NM_001145802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011583.mm.1 |
NM_001145803.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011583.mm.1 |
NM_001145803.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011583.mm.1 |
NM_008547.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011583.mm.1 |
NM_008547.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011582.mm.1 |
TC1300001857.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300011582.mm.1 |
NM_001145802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011582.mm.1 |
NM_001145802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011582.mm.1 |
NM_001145803.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011582.mm.1 |
NM_001145803.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011582.mm.1 |
NM_008547.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300011582.mm.1 |
NM_008547.2 |
Refseq |