Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01340560_s1
          See other SMARCA5-PS GE Assays ›
          Gene Symbol:
          Smarca5-ps
          Gene Name:
          SWI/SNF related, matrix associated, actin depenent ragulator of chromatin, subfamily a, member 5, pseudogene
          Gene Aliases:
          9230115A19Rik, OTTMUSG00000011029
          Chromosome Location:
          Chr.4: 145464210 - 145467961 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01340560_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smarca5-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002888.2 1 3369 190

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream tumor necrosis factor receptor superfamily,
          member 8
          Tnfrsf8 Cd30,
          D1S166E,
          Ki,
          Ki-1
          -149.1kb Anti-sense Chr. 4 - 145268976 - 145315147 on NCBI Build GRCm38
          Downstream predicted gene 13212 Gm13212 OTTMUSG00000010965 117.2kb Sense Chr. 4 - 145585172 - 145625345 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001863.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P462125 AK041999.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P462125 AK079035.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P462125 NR_002888.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P462125 AK041999.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P462125 AK079035.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P462125 NR_002888.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964559 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966525 NR_002888.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P462125 AK041999.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P462125 AK079035.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P462125 NR_002888.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P462125 AK041999.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P462125 AK079035.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P462125 NR_002888.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964559 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966525 NR_002888.2 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10510143 - -
          Affymetrix Mouse Gene 1.0 ST Array 10510177 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518363 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518362 - -
          Affymetrix Mouse Gene 1.1 ST Array 10510176 - -
          Affymetrix Mouse Gene 1.1 ST Array 10510142 - -
          Affymetrix Mouse Gene 1.1 ST Array 10518361 - -
          Affymetrix Mouse Clariom S Array PSR0400015469.mm.1 TC0400001863.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015469.mm.1 NR_002888.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015470.mm.1 TC0400001863.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015470.mm.1 NR_002888.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015471.mm.1 TC0400001863.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015471.mm.1 NR_002888.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015472.mm.1 TC0400001863.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015472.mm.1 NR_002888.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015473.mm.1 TC0400001863.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015473.mm.1 NR_002888.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015474.mm.1 TC0400001863.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015474.mm.1 NR_002888.2 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2616801 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225020 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2616801 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0