Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01341687_g1
          See other SMAD5 GE Assays ›
          Gene Symbol:
          Smad5
          Gene Name:
          SMAD family member 5
          Gene Aliases:
          1110051M15Rik, AI451355, Dwf-C, Madh5, MusMLP
          Chromosome Location:
          Chr.13: 56702988 - 56742379 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01341687_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smad5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164041.1 NP_001157513.1 5 - 6 1682 98
          NM_001164042.1 NP_001157514.1 5 - 6 1302 98
          NM_008541.3 NP_032567.1 5 - 6 1279 98
          GenBank mRNA AF063006.1 - 5 - 6 1683 98
          AK082997.1 - 5 - 6 1303 98
          BC050001.1 - 5 - 6 1182 6490853 98
          U58993.1 - 5 - 6 1258 98
          U77638.1 - 5 - 6 1428 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000694.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smad5

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          Mullerian duct regression
          osteoblast fate commitment
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein phosphorylation
          transforming growth factor beta receptor signaling pathway
          multicellular organism development
          germ cell development
          cell differentiation
          erythrocyte differentiation
          BMP signaling pathway
          positive regulation of osteoblast differentiation
          positive regulation of transcription by RNA polymerase II
          cartilage development
          cardiac muscle contraction
          bone development
          cellular response to organic cyclic compound
          cellular response to BMP stimulus
          positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
          RNA polymerase II core promoter sequence-specific DNA binding
          DNA binding
          DNA-binding transcription factor activity
          protein binding
          ubiquitin protein ligase binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300004616.mm.1 TC1300000694.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300004616.mm.1 NM_001164041.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004616.mm.1 NM_001164041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004616.mm.1 NM_001164042.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004616.mm.1 NM_001164042.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004616.mm.1 NM_008541.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004616.mm.1 NM_008541.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004618.mm.1 TC1300000694.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300004618.mm.1 NM_001164041.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004618.mm.1 NM_001164041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004618.mm.1 NM_001164042.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004618.mm.1 NM_001164042.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004618.mm.1 NM_008541.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004618.mm.1 NM_008541.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004612.mm.1 TC1300000694.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300004612.mm.1 NM_001164042.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004612.mm.1 NM_001164042.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004613.mm.1 TC1300000694.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300004613.mm.1 NM_001164042.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004613.mm.1 NM_001164042.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004623.mm.1 TC1300000694.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300004623.mm.1 NM_001164041.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004623.mm.1 NM_001164041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004623.mm.1 NM_001164042.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004623.mm.1 NM_001164042.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004623.mm.1 NM_008541.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004623.mm.1 NM_008541.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004605.mm.1 TC1300000694.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300004605.mm.1 NM_008541.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004605.mm.1 NM_008541.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004619.mm.1 TC1300000694.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300004619.mm.1 NM_001164041.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004619.mm.1 NM_001164041.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004619.mm.1 NM_001164042.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004619.mm.1 NM_001164042.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004619.mm.1 NM_008541.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300004619.mm.1 NM_008541.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0