Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172520.2 | NP_766108.1 | 15 - 16 | 2448 | 73 | |
GenBank mRNA | AK031443.1 | - | 15 - 16 | 2449 | 73 | |
AK053075.1 | - | 15 - 16 | 2449 | 73 |
Set Membership: |
![]() ![]() ![]() ![]() |