Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01343129_m1
          See other SENP2 GE Assays ›
          Gene Symbol:
          Senp2
          Gene Name:
          SUMO/sentrin specific peptidase 2
          Gene Aliases:
          2310007L05Rik, 4930538C18Rik, AI646780, AW554757, Smt3ip2, SuPr-1, mKIAA1331
          Chromosome Location:
          Chr.16: 22009432 - 22049269 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01343129_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Senp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029457.3 NP_083733.1 13 - 14 1598 150
          NR_027488.1 13 - 14 1665 150
          GenBank mRNA AF368904.1 - 13 - 14 1644 150
          AK015987.1 - 13 - 14 1577 150
          AK031030.1 - 13 - 14 1583 150
          AY188288.1 - 12 - 13 1370 150
          BC031652.1 - 13 - 14 1573 3483046 150
          BC040805.1 - 5 - 6 1542 150
          BC042514.1 - 5 - 6 1542 150

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000307.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Senp2

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          proteolysis
          transport
          heart development
          dorsal/ventral axis specification
          protein transport
          Wnt signaling pathway
          protein desumoylation
          negative regulation of protein ubiquitination
          positive regulation of protein ubiquitination
          protein destabilization
          negative regulation of protein binding
          regulation of DNA endoreduplication
          negative regulation of chromatin binding
          negative regulation of DNA damage response, signal transduction by p53 class mediator
          fat cell differentiation
          positive regulation of transcription by RNA polymerase II
          mRNA transport
          regulation of protein metabolic process
          trophoblast giant cell differentiation
          labyrinthine layer development
          spongiotrophoblast layer development
          negative regulation of signal transduction by p53 class mediator
          regulation of G1/S transition of mitotic cell cycle
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity
          SUMO-specific protease activity
          protein domain specific binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1600003132.mm.1 TC1600000307.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003132.mm.1 NM_029457.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003132.mm.1 NM_029457.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003132.mm.1 NR_027488.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003132.mm.1 NR_027488.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003129.mm.1 TC1600000307.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003129.mm.1 NM_029457.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003129.mm.1 NM_029457.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003129.mm.1 NR_027488.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003129.mm.1 NR_027488.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003126.mm.1 TC1600000307.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003126.mm.1 NM_029457.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003126.mm.1 NM_029457.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003126.mm.1 NR_027488.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003126.mm.1 NR_027488.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003127.mm.1 TC1600000307.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003127.mm.1 NM_029457.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003127.mm.1 NM_029457.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003127.mm.1 NR_027488.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003127.mm.1 NR_027488.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003131.mm.1 TC1600000307.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003131.mm.1 NM_029457.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003131.mm.1 NM_029457.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003131.mm.1 NR_027488.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003131.mm.1 NR_027488.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003130.mm.1 TC1600000307.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003130.mm.1 NM_029457.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003130.mm.1 NM_029457.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003130.mm.1 NR_027488.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003130.mm.1 NR_027488.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0