Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01344212_m1
          See other TMEM30C GE Assays ›
          Gene Symbol:
          Tmem30c
          Gene Name:
          transmembrane protein 30C
          Gene Aliases:
          4933401B01Rik, 4933409A18Rik, Cdc50c
          Chromosome Location:
          Chr.16: 57266138 - 57301893 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01344212_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem30c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027651.1 NP_081927.1 7 - 8 921 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001760.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957983 NM_027651.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957983 AK016597.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2192497 AK016597.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 108029 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 878531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 486708 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359193 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524846 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 347340 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 645074 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 230503 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 859422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 965083 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 283873 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 14615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 174026 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167710 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180254 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131718 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142715 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957983 NM_027651.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957983 AK016597.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2192497 AK016597.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P180254 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131718 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142715 - -
          Whole Mouse Genome G4122A A_52_P198561 AK161475.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P198561 NM_027651.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P198561 AK016597.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56447 AK161475.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56447 AK016747.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P447540 NM_027651.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P447540 AK016597.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142715 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P180254 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13015193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12729444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12516758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12674489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12516764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30785194 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10440074 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440075 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440073 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440076 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440071 - -
          Affymetrix Mouse Gene 1.0 ST Array 10440072 - -
          Affymetrix HT MG-430 PM Array Plate 1431455_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1454536_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1433275_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10440070 - -
          Mouse Genome 430 2.0 Array 1433275_PM_at - -
          Mouse Genome 430 2.0 Array 1454536_at - -
          Mouse Genome 430 2.0 Array 1431455_PM_at - -
          Mouse Genome 430 2.0 Array 1431455_at - -
          Mouse Genome 430 2.0 Array 1433275_at - -
          Mouse Genome 430 2.0 Array 1454536_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71027_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71027_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600014305.mm.1 TC1600001760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600014305.mm.1 NM_027651.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014305.mm.1 NM_027651.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014302.mm.1 TC1600001760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600014302.mm.1 NM_027651.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014302.mm.1 NM_027651.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014303.mm.1 TC1600001760.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600014303.mm.1 NM_027651.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600014303.mm.1 NM_027651.1 Refseq
          Mouse Expression Set 430 1431455_at - -
          Mouse Expression Set 430 1431455_PM_at - -
          Murine Genome U74v2 Set 167393_r_at - -
          Murine Genome U74v2 Set 167393_PM_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1454536_at - -
          Affymetrix HT Mouse Genome 430B Array 1433275_at - -
          Affymetrix HT Mouse Genome 430B Array 1431455_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433275_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431455_at - -
          Affymetrix HT Mouse Genome 430B Array 1454536_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 406463 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2651251 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2651251 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0