Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01344684_m1
          See other DTX3L GE Assays ›
          Gene Symbol:
          Dtx3l
          Gene Name:
          deltex 3-like (Drosophila)
          Gene Aliases:
          AU042200, BC023741
          Chromosome Location:
          Chr.16: 35926515 - 35939027 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01344684_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dtx3l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013371.2 NP_001013389.2 4 - 5 2327 107
          GenBank mRNA AK146802.1 - 4 - 5 2235 107
          BC023741.1 - 2 - 3 1129 107
          BC025947.1 - 2 - 3 411 107
          BC120494.1 - 4 - 5 2268 107
          BC137694.1 - 4 - 5 2268 8860713 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001557.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dtx3l

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          double-strand break repair
          chromatin organization
          cellular response to DNA damage stimulus
          histone monoubiquitination
          ubiquitin-protein transferase activity
          zinc ion binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          histone binding
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          metal ion binding
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30672895 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30672897 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30672902 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30672899 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30672896 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30672898 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30672900 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174541 BC137694.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174541 BC023741.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174541 BC085093.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174541 BC120494.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174541 AK146802.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059154 BC085093.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2059154 NM_001013371.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P106060 AK090152.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 802319 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749490 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352899 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 772975 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430045 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 150413 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 469697 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 355963 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628595 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 112317 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141062 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174541 BC137694.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174541 BC023741.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174541 BC085093.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174541 BC120494.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174541 AK146802.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059154 BC085093.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2059154 NM_001013371.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P106060 AK090152.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30672910 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141062 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03169866 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03169866 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141062 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12529406 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12529396 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12529404 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12529405 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12529393 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30672899 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30672895 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30672897 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30672896 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30672900 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30672898 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30672902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30672938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12747375 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12707481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12672641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12723884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12474102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12791703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12970333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12529405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672928 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12723882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30672902 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10439271 - -
          Affymetrix Mouse Gene 1.0 ST Array 10439274 - -
          Affymetrix Mouse Gene 1.0 ST Array 10439269 - -
          Affymetrix Mouse Gene 1.0 ST Array 10439275 - -
          Affymetrix Mouse Gene 1.0 ST Array 10439273 - -
          Affymetrix Mouse Gene 1.0 ST Array 10439270 - -
          Affymetrix HT MG-430 PM Array Plate 1435208_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1439825_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10435457 - -
          Affymetrix Mouse Gene 1.1 ST Array 10439268 - -
          Mouse Genome 430 2.0 Array 1435208_at - -
          Mouse Genome 430 2.0 Array 1435208_PM_at - -
          Mouse Genome 430 2.0 Array 1439825_PM_at - -
          Mouse Genome 430 2.0 Array 1439825_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 209200_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 209200_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600012774.mm.1 TC1600001557.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012774.mm.1 NM_001013371.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012774.mm.1 NM_001013371.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012775.mm.1 TC1600001557.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600012775.mm.1 NM_001013371.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600012775.mm.1 NM_001013371.2 Refseq
          Mouse Expression Set 430 1435208_at - -
          Mouse Expression Set 430 1439825_at - -
          Mouse Expression Set 430 1439825_PM_at - -
          Mouse Expression Set 430 1435208_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435208_at - -
          Affymetrix HT Mouse Genome 430B Array 1435208_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439825_at - -
          Affymetrix HT Mouse Genome 430B Array 1439825_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 783681 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100520377 - -
          Illumina mouse-6 v1.0 expression beadchip 100520377 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220813 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220813 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247917 - -
          Illumina Sentrix mouse-6.v1 bead array 100520377 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100520377 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247917 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix