Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix HT MG-430 PM Array Plate |
1448099_PM_at |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1439560_PM_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1439560_x_at |
- |
- |
Mouse Genome 430 2.0 Array |
1448099_at |
- |
- |
Mouse Genome 430 2.0 Array |
1448099_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1439560_PM_x_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
432995_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
432995_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1600000398.mm.1 |
TC1600000043.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000398.mm.1 |
NM_001253796.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000398.mm.1 |
NM_001253796.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000398.mm.1 |
NM_001253797.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000398.mm.1 |
NM_001253797.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000398.mm.1 |
NM_001253802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000398.mm.1 |
NM_001253802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000398.mm.1 |
NM_001253803.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000398.mm.1 |
NM_001253803.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000398.mm.1 |
NR_045595.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000398.mm.1 |
NR_045595.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000398.mm.1 |
NR_045596.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000398.mm.1 |
NR_045596.1 |
Refseq |
Mouse Expression Set 430 |
1448099_at |
- |
- |
Mouse Expression Set 430 |
1439560_PM_x_at |
- |
- |
Mouse Expression Set 430 |
1448099_PM_at |
- |
- |
Mouse Expression Set 430 |
1439560_x_at |
- |
- |
Murine Genome U74v2 Set |
165927_at |
- |
- |
Murine Genome U74v2 Set |
165927_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1439560_PM_x_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1439560_x_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1448099_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1448099_at |
- |
- |