Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01347455_g1
          See other 4930547H16RIK GE Assays ›
          Gene Symbol:
          4930547H16Rik
          Gene Name:
          RIKEN cDNA 4930547H16 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 69771890 - 69772714 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01347455_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930547H16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK016056.1 - 1 - 2 210 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ubiquitin-conjugating enzyme E2Q family-like 1 Ube2ql1 3110006E14Rik -31.6kb Sense Chr. 13 - 69702832 - 69739897 on NCBI Build GRCm38
          Downstream mediator complex subunit 10 Med10 AA959813,
          AI385605,
          C78613,
          D13Wsu50e
          29.7kb Anti-sense Chr. 13 - 69802348 - 69816104 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957178 AK016056.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 269588 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957178 AK016056.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10673992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29117688 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 449420 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0