Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01347901_g1
          See other PPBP GE Assays ›
          Gene Symbol:
          Ppbp
          Gene Name:
          pro-platelet basic protein
          Gene Aliases:
          2400003M24Rik, AI854500, CTAP3, CTAPIII, Cxcl7, LA-PF4, LDGF, MDGF, NAP-2, NAP-2-L1, Scyb7, TGB, TGB1, THBGB1, b-TG1, beta-TG
          Chromosome Location:
          Chr.5: 90768518 - 90770060 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01347901_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppbp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023785.2 NP_076274.1 2 - 3 341 67
          GenBank mRNA AB042817.1 - 2 - 3 350 67
          AF219112.1 - 2 - 3 341 67
          AF278700.1 - 2 - 3 341 67
          AK010273.1 - 2 - 3 342 67
          AK157077.1 - 2 - 3 341 67
          AK157141.1 - 2 - 3 342 67
          AK158061.1 - 2 - 3 339 67
          AK171720.1 - 2 - 3 342 67
          BC127043.1 - 2 - 3 308 40053104 67
          BC127044.1 - 2 - 3 308 40053105 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000842.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppbp

          Gene Ontology Categories:

          Function(s) Process(es)

          leukocyte migration involved in inflammatory response
          positive regulation of leukocyte chemotaxis
          inflammatory response
          immune response
          G protein-coupled receptor signaling pathway
          response to lipopolysaccharide
          regulation of cell population proliferation
          cell chemotaxis
          chemokine-mediated signaling pathway
          chemokine activity
          CXCR chemokine receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P428372 NM_023785.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P428372 AF219112.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P428372 NM_023785.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P428372 AF219112.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 106461 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174831 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P428372 NM_023785.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P428372 AF219112.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P428372 NM_023785.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P428372 AF219112.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P428372 NM_023785.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P428372 AF219112.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P428372 NM_023785.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P428372 AF219112.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23885061 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174831 - -
          Whole Mouse Genome G4122A A_51_P428372 NM_023785.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P428372 AF219112.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P428372 NM_023785.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P428372 AF219112.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105592 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174831 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04182535 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04182545 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04182537 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04182533 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04182532 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04182530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23885102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04836485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04878220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04182533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04443131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885075 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04836500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885101 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04311772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23885104 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10523129 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523130 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523133 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523131 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523132 - -
          Affymetrix HT MG-430 PM Array Plate 1418480_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10523128 - -
          Mouse Genome 430 2.0 Array 1418480_at - -
          Mouse Genome 430 2.0 Array 1418480_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57349_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57349_at - -
          Affymetrix Mouse Clariom S Array PSR0500007415.mm.1 TC0500000842.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007415.mm.1 NM_023785.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007416.mm.1 TC0500000842.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007416.mm.1 NM_023785.3 Refseq
          Mouse Expression Set 430 1418480_at - -
          Mouse Expression Set 430 1418480_PM_at - -
          Mouse Genome 430A 2.0 Array 1418480_PM_at - -
          Mouse Genome 430A 2.0 Array 1418480_at - -
          Affymetrix HT Mouse Genome 430A Array 1418480_at - -
          Affymetrix HT Mouse Genome 430A Array 1418480_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 767832 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5130446 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005130446 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228102 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228102 - -
          mouse-6 v1.1 (Illumina) scl27603.2.1_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 5130446 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27603.2.1_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5130446 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228102 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2908435 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2908435 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228102 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl27603.2.1_3-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0