Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01348977_m1
          See other BC030870 GE Assays ›
          Gene Symbol:
          BC030870
          Gene Name:
          cDNA sequence BC030870
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 65085613 - 65129537 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01348977_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BC030870
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033217.1 2 - 3 431 141
          GenBank mRNA AK018670.1 - 2 - 3 218 141
          AK131860.1 - 2 - 3 456 141
          AK136454.1 - 2 - 3 306 141
          BC030870.1 - 2 - 3 152 141

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002322.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126348 AK136454.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126348 BC030870.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126348 AK018670.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126348 NR_033217.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165787 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06537814 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06537823 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01755659 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165787 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06537789 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126348 AK136454.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126348 BC030870.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126348 AK018670.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126348 NR_033217.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07136047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06898040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07281893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07087478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07159270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07410434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927302 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927333 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06765999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06747459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06669656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07136056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07152563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07281905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07136044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07369405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07371985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927311 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06669663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07281901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25927162 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1459904_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440094_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10578952 - -
          Mouse Genome 430 2.0 Array 1440094_at - -
          Mouse Genome 430 2.0 Array 1459904_PM_at - -
          Mouse Genome 430 2.0 Array 1440094_PM_at - -
          Mouse Genome 430 2.0 Array 1459904_at - -
          Affymetrix Mouse Clariom S Array PSR0800017529.mm.1 TC0800002322.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017529.mm.1 NR_033217.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017529.mm.1 NR_033217.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017528.mm.1 TC0800002322.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800017528.mm.1 NR_033217.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800017528.mm.1 NR_033217.1 Refseq
          Mouse Expression Set 430 1440094_at - -
          Mouse Expression Set 430 1440094_PM_at - -
          Mouse Expression Set 430 1459904_at - -
          Mouse Expression Set 430 1459904_PM_at - -
          Murine Genome U74v2 Set 131164_PM_at - -
          Murine Genome U74v2 Set 167289_at - -
          Murine Genome U74v2 Set 131164_at - -
          Murine Genome U74v2 Set 167289_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459904_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440094_at - -
          Affymetrix HT Mouse Genome 430B Array 1459904_at - -
          Affymetrix HT Mouse Genome 430B Array 1440094_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 885992 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0