Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800017225.mm.1 |
TC0800002265.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017225.mm.1 |
NM_175032.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017225.mm.1 |
NM_175032.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017234.mm.1 |
TC0800002265.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017234.mm.1 |
NM_175032.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017234.mm.1 |
NM_175032.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017223.mm.1 |
TC0800002265.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017223.mm.1 |
NM_175032.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017223.mm.1 |
NM_175032.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017235.mm.1 |
TC0800002265.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017235.mm.1 |
NM_175032.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017235.mm.1 |
NM_175032.3 |
Refseq |