Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400020540.mm.1 |
TC0400002527.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020540.mm.1 |
NM_001013377.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020540.mm.1 |
NM_001013377.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020547.mm.1 |
TC0400002527.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020547.mm.1 |
NM_001013377.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020547.mm.1 |
NM_001013377.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020531.mm.1 |
TC0400002527.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020531.mm.1 |
NM_001013377.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020531.mm.1 |
NM_001013377.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020539.mm.1 |
TC0400002527.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020539.mm.1 |
NM_001013377.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020539.mm.1 |
NM_001013377.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020534.mm.1 |
TC0400002527.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020534.mm.1 |
NM_001013377.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020534.mm.1 |
NM_001013377.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020536.mm.1 |
TC0400002527.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020536.mm.1 |
NM_001013377.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020536.mm.1 |
NM_001013377.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020530.mm.1 |
TC0400002527.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020530.mm.1 |
NM_001013377.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020530.mm.1 |
NM_001013377.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020543.mm.1 |
TC0400002527.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400020543.mm.1 |
NM_001013377.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400020543.mm.1 |
NM_001013377.2 |
Refseq |