Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01350990_m1
          See other 4930515B02RIK GE Assays ›
          Gene Symbol:
          4930515B02Rik
          Gene Name:
          RIKEN cDNA 4930515B02 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 140872693 - 140878057 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01350990_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930515B02Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040654.1 1 - 2 173 70
          GenBank mRNA AK015793.1 - 1 - 2 173 70
          BY715338.1 - 1 - 2 171 70

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream peptidyl arginine deiminase,
          type I
          Padi1 AV236283,
          Pdi1
          -26.9kb Anti-sense Chr. 4 - 140812981 - 140845778 on NCBI Build GRCm38
          Downstream peptidyl arginine deiminase,
          type II
          Padi2 Pdi,
          Pdi2,
          mKIAA0994
          28.6kb Sense Chr. 4 - 140906334 - 140952586 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001769.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726298 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380621 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896597 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05428948 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139188 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23360775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03335280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23360766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03335288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23360758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23360734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03264891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03645970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23360814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03264902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23360733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23360812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03264898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03690591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23360824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04085921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03264889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23360763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23360737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03376596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03335287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03264903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03658502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23360822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03252977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23360765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23360813 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400014581.mm.1 TC0400001769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014581.mm.1 NR_040654.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014581.mm.1 NR_040654.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014583.mm.1 TC0400001769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014583.mm.1 NR_040654.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014583.mm.1 NR_040654.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014585.mm.1 TC0400001769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014585.mm.1 NR_040654.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014585.mm.1 NR_040654.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014584.mm.1 TC0400001769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014584.mm.1 NR_040654.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014584.mm.1 NR_040654.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014586.mm.1 TC0400001769.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014586.mm.1 NR_040654.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014586.mm.1 NR_040654.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 702467 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100840563 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0