Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01351183_m1
          See other 4930512H18RIK GE Assays ›
          Gene Symbol:
          4930512H18Rik
          Gene Name:
          RIKEN cDNA 4930512H18 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 61913060 - 61924768 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01351183_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930512H18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK084432.1 - 223 67
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB658464.1 - 2 - 3 223 Chr. 8 - 61849006 - 61921991 [+] on Build GRCm38 67

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          35254
          Gm35254 -218.5kb Sense Chr. 8 - 61620404 - 61632929 on NCBI Build GRCm38
          Downstream DEAD (Asp-Glu-Ala-Asp) box polypeptide 60 Ddx60 9830118M07 6.1kb Sense Chr. 8 - 61928066 - 62037704 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2305560 AK084432.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460560 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 918069 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 387007 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 32018 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485910 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790300 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 589599 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 87599 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 797399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 189534 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 660238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 585603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139829 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16331 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158344 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112120 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149718 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119623 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2305560 AK084432.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158344 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06531650 - -
          Whole Mouse Genome G4122A A_52_P242397 AK084432.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P242397 AK015772.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137897 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25912957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25912948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25912940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06779334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25912955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25912943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25912946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25912953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25912939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25912949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25912951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25912950 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1458115_PM_at - -
          Mouse Genome 430 2.0 Array 1458115_at - -
          Mouse Genome 430 2.0 Array 1458115_PM_at - -
          Mouse Expression Set 430 1458115_PM_at - -
          Mouse Expression Set 430 1458115_at - -
          Affymetrix HT Mouse Genome 430B Array 1458115_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458115_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105360358 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline