Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100005543.mm.1 |
TC1100000689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100005543.mm.1 |
NM_001083884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005543.mm.1 |
NM_001083884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005543.mm.1 |
NM_027339.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005543.mm.1 |
NM_027339.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005544.mm.1 |
TC1100000689.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100005544.mm.1 |
NM_001083884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100005544.mm.1 |
NM_001083884.1 |
Refseq |