Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01352299_g1
          See other ISYNA1 GE Assays ›
          Gene Symbol:
          Isyna1
          Gene Name:
          myo-inositol 1-phosphate synthase A1
          Gene Aliases:
          1300017C10Rik, AU018670
          Chromosome Location:
          Chr.8: 70594481 - 70597290 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01352299_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Isyna1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023627.1 NP_076116.1 10 - 11 1546 141
          GenBank mRNA AF288525.1 - 10 - 11 1546 141
          AK005029.2 - 10 - 11 1524 141
          AK079323.1 - 10 - 11 1523 141
          AK137089.1 - 9 - 10 1628 141
          BC003458.1 - 10 - 11 1537 2648362 141

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000756.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Isyna1

          Gene Ontology Categories:

          Function(s) Process(es)

          inositol biosynthetic process
          lipid metabolic process
          biological_process
          phospholipid biosynthetic process
          inositol-3-phosphate synthase activity
          isomerase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952862 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952864 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952871 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952869 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952872 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952865 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952866 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952861 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952863 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952873 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952870 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P228658 BC003458.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P228658 AK005029.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P228658 AK079323.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P228658 AK137089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P228658 NM_023627.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P228658 BC003458.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P228658 AK005029.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P228658 AK079323.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P228658 AK137089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P228658 NM_023627.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 645866 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 131897 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134806 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148986 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P228658 BC003458.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P228658 AK005029.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P228658 AK079323.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P228658 AK137089.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P228658 NM_023627.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P228658 BC003458.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P228658 AK005029.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P228658 AK079323.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P228658 AK137089.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P228658 NM_023627.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P228658 BC003458.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P228658 AK005029.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P228658 AK079323.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P228658 AK137089.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P228658 NM_023627.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P228658 BC003458.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P228658 AK005029.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P228658 AK079323.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P228658 AK137089.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P228658 NM_023627.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25952859 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148986 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134806 - -
          Whole Mouse Genome G4122A A_51_P228658 BC003458.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P228658 AK005029.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P228658 AK079323.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P228658 AK137089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P228658 NM_023627.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P228658 BC003458.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P228658 AK005029.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P228658 AK079323.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P228658 AK137089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P228658 NM_023627.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139512 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148986 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134806 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07440816 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07440811 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07440808 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07440813 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07440807 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07440815 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952861 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952866 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952865 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952870 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952862 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952872 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952864 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952869 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952871 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952863 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25952873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25952957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25952856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25952876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25952887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25952859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07230915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25952851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25952883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25952852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07230913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06937115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07230921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25952884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25952864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07083157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25952865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25952881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25952857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25952863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07440813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07230907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25952877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07230923 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10572435 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572440 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572433 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572436 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572442 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572441 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572444 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572438 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572434 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572443 - -
          Affymetrix Mouse Gene 1.0 ST Array 10572439 - -
          Affymetrix HT MG-430 PM Array Plate 1415977_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10572432 - -
          Mouse Genome 430 2.0 Array 1415977_PM_at - -
          Mouse Genome 430 2.0 Array 1415977_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71780_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71780_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800005369.mm.1 TC0800000756.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005369.mm.1 NM_023627.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005369.mm.1 NM_023627.1 Refseq
          Mouse Expression Set 430 1415977_at - -
          Mouse Expression Set 430 1415977_PM_at - -
          Murine Genome U74v2 Set 162401_f_at - -
          Murine Genome U74v2 Set 162401_PM_f_at - -
          Murine Genome U74v2 Set 160337_PM_at - -
          Murine Genome U74v2 Set 160337_at - -
          Mouse Genome 430A 2.0 Array 1415977_PM_at - -
          Mouse Genome 430A 2.0 Array 1415977_at - -
          Affymetrix HT Mouse Genome 430A Array 1415977_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1415977_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1170692 - -
          Illumina mouse-6 v1.0 expression beadchip 1170692 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001170692 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228752 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228752 - -
          mouse-6 v1.1 (Illumina) scl071780.11_24-S - -
          Illumina Sentrix mouse-6.v1 bead array 1170692 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl071780.11_24-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170692 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228752 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228752 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0