Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01354089_g1
          See other FAM206A GE Assays ›
          Gene Symbol:
          Fam206a
          Gene Name:
          family with sequence similarity 206, member A
          Gene Aliases:
          simiate
          Chromosome Location:
          Chr.4: 56802329 - 56809605 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01354089_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam206a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081420.1 NP_001074889.1 5 - 6 732 132
          GenBank mRNA AK052647.1 - 5 - 6 547 132
          AK164596.1 - 5 - 6 545 132
          BC026590.1 - 5 - 6 711 4973099 132
          BC048548.1 - 5 - 6 683 6590165 132
          BC062190.1 - 6 - 7 713 132

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000622.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1971639 NM_001081420.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 684468 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973049 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777741 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633779 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 109212 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165022 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1971639 NM_001081420.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165022 - -
          Whole Mouse Genome G4122A A_51_P326566 BC062190.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326566 AK164596.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P326566 AK043588.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660713 BC026590.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660713 BC048548.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660713 BC062190.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660713 AK052647.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P660713 AK164596.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118630 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165022 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03405593 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03405597 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03405590 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03405592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22941582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03867425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03333973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03590023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03787294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03590018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03787300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03590027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22941599 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10505115 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505110 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505114 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505112 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505116 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505113 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505117 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505111 - -
          Affymetrix HT MG-430 PM Array Plate 1436422_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10505109 - -
          Mouse Genome 430 2.0 Array 1436422_PM_at - -
          Mouse Genome 430 2.0 Array 1436422_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 230234_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 230234_at - -
          Affymetrix Mouse Clariom S Array PSR0400004449.mm.1 TC0400000622.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400004449.mm.1 NM_001081420.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004449.mm.1 NM_001081420.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004450.mm.1 TC0400000622.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400004450.mm.1 NM_001081420.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004450.mm.1 NM_001081420.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004448.mm.1 TC0400000622.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400004448.mm.1 NM_001081420.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004448.mm.1 NM_001081420.1 Refseq
          Mouse Expression Set 430 1436422_PM_at - -
          Mouse Expression Set 430 1436422_at - -
          Murine Genome U74v2 Set 130572_PM_at - -
          Murine Genome U74v2 Set 137550_at - -
          Murine Genome U74v2 Set 137550_PM_at - -
          Murine Genome U74v2 Set 130572_at - -
          Affymetrix HT Mouse Genome 430B Array 1436422_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436422_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257772 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2734007 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2638690 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3051252 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1257772 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3051252 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0