Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00019907.mm.1 |
TC0X00003168.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019907.mm.1 |
NM_001199246.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019907.mm.1 |
NM_001199246.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019907.mm.1 |
NM_030700.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019907.mm.1 |
NM_030700.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019908.mm.1 |
TC0X00003168.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019908.mm.1 |
NM_001199246.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019908.mm.1 |
NM_001199246.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019908.mm.1 |
NM_030700.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019908.mm.1 |
NM_030700.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019919.mm.1 |
TC0X00003168.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019919.mm.1 |
NM_001199246.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019919.mm.1 |
NM_001199246.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019909.mm.1 |
TC0X00003168.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019909.mm.1 |
NM_001199246.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019909.mm.1 |
NM_001199246.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019909.mm.1 |
NM_030700.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019909.mm.1 |
NM_030700.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019920.mm.1 |
TC0X00003168.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019920.mm.1 |
NM_001199246.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019920.mm.1 |
NM_001199246.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019911.mm.1 |
TC0X00003168.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019911.mm.1 |
NM_001199246.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019911.mm.1 |
NM_001199246.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019911.mm.1 |
NM_030700.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019911.mm.1 |
NM_030700.2 |
Refseq |