Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01612271_m1
          See other FAM177A GE Assays ›
          Gene Symbol:
          Fam177a 1700047I17Rik2
          Gene Name:
          family with sequence similarity 177, member A
          RIKEN cDNA 1700047I17 gene 2
          Gene Aliases:
          1700047I17Rik, 1700047I17Rik1, C14orf24, D730001C10Rik, Fam177a1
          -
          Chromosome Location:
          Chr.12: 55124468 - 55142104 on Build GRCm38
          Chromosome Location:
          Chr.12: 55199532 - 55217168 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm01612271_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam177a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028527.1 NP_082803.1 3 - 4 441 60
          GenBank mRNA AK006713.1 - 274 60
          AK045530.1 - 501 60
          AK159923.1 - 479 60
          BC048158.1 - 441 5698044 60
          BC049669.1 - 500 6535412 60
          Gene Symbol: 1700047I17Rik2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100116.1 NP_001093586.1 441 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000459.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam177a
          Click here to find siRNAs for Gene 1700047I17Rik2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28450733 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28450735 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28450736 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28450734 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28450732 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114269 NM_001100116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114269 BC048158.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114269 AK052876.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114269 NM_028527.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2449750 NM_001100116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2449750 BC048158.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2449750 AK045530.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2449750 NM_028527.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2449750 NM_001100116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2449750 BC048158.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2449750 AK045530.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2449750 NM_028527.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114269 NM_001100116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114269 BC048158.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114269 AK052876.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114269 NM_028527.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28450732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28450735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28450737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28450727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28450746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28450745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28450751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28450731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28450748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28450743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28450733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28450738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28450724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28450741 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172729 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07627006 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139974 - -
          Whole Mouse Genome G4122A A_52_P252996 NM_001100116.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P252996 BC048158.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P252996 AK045530.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P252996 AK052876.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P252996 NM_028527.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P252305 NM_001100116.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P252305 AK146826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P252305 BC048158.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P252305 AK045530.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P252305 AK159923.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172729 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139974 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10368158 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10373589 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10373586 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10373588 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28450733 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28450732 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28450735 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28450736 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28450734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28450783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10368157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10373589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450775 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10423867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10324518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10189944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09923914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10182052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10327732 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10368152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09962997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10168338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10182051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450773 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28450752 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10395774 - -
          Affymetrix Mouse Gene 1.0 ST Array 10400394 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395783 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395804 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395782 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395775 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395777 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395806 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395786 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395771 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395773 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395772 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395785 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395787 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395776 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395803 - -
          Affymetrix Mouse Gene 1.0 ST Array 10395781 - -
          Affymetrix Mouse Gene 1.1 ST Array 10400393 - -
          Affymetrix Mouse Gene 1.1 ST Array 10395805 - -
          Affymetrix Mouse Gene 1.1 ST Array 10395780 - -
          Affymetrix Mouse Gene 1.1 ST Array 10395770 - -
          Mouse Genome 430 2.0 Array 1460466_at - -
          Mouse Genome 430 2.0 Array 1445334_at - -
          Mouse Genome 430 2.0 Array 1445334_PM_at - -
          Mouse Genome 430 2.0 Array 1431268_PM_at - -
          Mouse Genome 430 2.0 Array 1460466_PM_at - -
          Mouse Genome 430 2.0 Array 1431268_at - -
          Affymetrix Mouse Clariom S Array PSR1200003145.mm.1 TC1200000459.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003145.mm.1 NM_001100116.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003145.mm.1 NM_028527.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003145.mm.1 NM_028527.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003144.mm.1 TC1200000459.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003144.mm.1 NM_001100116.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003144.mm.1 NM_028527.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003144.mm.1 NM_028527.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003143.mm.1 TC1200000459.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003143.mm.1 NM_001100116.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003143.mm.1 NM_028527.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003143.mm.1 NM_028527.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003142.mm.1 TC1200000459.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200003142.mm.1 NM_001100116.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003142.mm.1 NM_028527.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200003142.mm.1 NM_028527.1 Refseq
          Mouse Expression Set 430 1460466_PM_at - -
          Mouse Expression Set 430 1445334_PM_at - -
          Mouse Expression Set 430 1460466_at - -
          Mouse Expression Set 430 1460466_PM_at - -
          Mouse Expression Set 430 1445334_at - -
          Mouse Expression Set 430 1460466_at - -
          Affymetrix HT Mouse Genome 430B Array 1445334_at - -
          Affymetrix HT Mouse Genome 430B Array 1445334_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 455988 - -
          ABI Mouse Genome Survey Microarray 532846 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3190471 - -
          Illumina mouse-6 v1.0 expression beadchip 6770435 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003190471 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006770435 - -
          mouse-6 v1.1 (Illumina) scl073385.6_17-S - -
          mouse-6 v1.1 (Illumina) scl0073385.1_119-S - -
          Illumina Sentrix mouse-6.v1 bead array 3190471 - -
          Illumina Sentrix mouse-6.v1 bead array 6770435 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0073385.1_119-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl073385.6_17-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3190471 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6770435 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3107690 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3034034 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2730662 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3034034 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3107690 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229119 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0