Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01700367_g1
          See other PIRB GE Assays ›
          Gene Symbol:
          Pirb
          Gene Name:
          paired Ig-like receptor B
          Gene Aliases:
          Gp91, LIR-3, Lilrb3, PIR-B
          Chromosome Location:
          Chr.7: 3712505 - 3720456 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01700367_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pirb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011095.2 NP_035225.2 14 - 15 2405 71
          GenBank mRNA AK142375.1 - 14 - 15 2386 71
          AK152232.1 - 13 - 14 2608 71
          AK153116.1 - 13 - 14 2608 71
          AK161081.1 - 2376 71
          AK170720.1 - 11 - 12 3033 71
          BC026937.1 - 2359 4488338 71
          U83172.1 - 14 - 15 2349 71
          U96689.1 - 14 - 15 2388 71
          U96691.1 - 2352 71
          U96692.1 - 14 - 15 2389 71
          U96693.1 - 14 - 15 2405 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004631.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pirb

          Gene Ontology Categories:

          Function(s)

          B cell homeostasis
          immune system process
          cytokine-mediated signaling pathway
          B cell mediated immunity
          myeloid dendritic cell differentiation

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2097279 U83172.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2097279 AK161081.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2097279 AK142375.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2097279 NM_011095.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 937815 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 365306 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 929748 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 100040 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124622 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2097279 U83172.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2097279 AK161081.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2097279 AK142375.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2097279 NM_011095.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2053398 AK161081.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P445728 U96692.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P445728 U96693.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P445728 U96691.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P445728 U96689.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P445728 BC026937.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P445726 U96692.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P445726 U96693.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P445726 U96691.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P445726 U96689.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P445726 BC026937.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24947112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24947104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24947111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24947114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24947108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24947101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24947107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24947109 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03806327 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06112726 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06112707 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124622 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06112746 - -
          Whole Mouse Genome G4122A A_51_P445728 U96692.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P445728 U96693.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P445728 U96691.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P445728 U96689.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P445728 BC026937.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P445726 U96692.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P445726 U96693.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P445726 U96691.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P445726 U96689.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P445726 BC026937.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06112707 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124622 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06112726 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06112746 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03806327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24948379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32979800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946969 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10559448 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559449 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559461 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559469 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559459 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559462 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559458 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559472 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559475 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559447 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559464 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559456 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559465 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559470 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559477 - -
          Affymetrix HT MG-430 PM Array Plate 1424302_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10559446 - -
          Mouse Genome 430 2.0 Array 1424302_PM_at - -
          Mouse Genome 430 2.0 Array 1424302_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18733_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18733_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700019296.mm.1 TC0700004631.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019296.mm.1 NM_011095.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019296.mm.1 NM_011095.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019294.mm.1 TC0700004631.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019294.mm.1 NM_011095.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019294.mm.1 NM_011095.2 Refseq
          Mouse Expression Set 430 1420464_PM_s_at - -
          Mouse Expression Set 430 1420464_s_at - -
          Mouse Expression Set 430 1424302_PM_at - -
          Mouse Expression Set 430 1424302_at - -
          Murine Genome U74v2 Set 100328_s_at - -
          Murine Genome U74v2 Set 100328_PM_s_at - -
          Murine Genome U74v2 Set 98003_at - -
          Murine Genome U74v2 Set 98003_PM_at - -
          Murine Genome U74v2 Set 168294_f_at - -
          Murine Genome U74v2 Set 168294_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1424302_at - -
          Mouse Genome 430A 2.0 Array 1420464_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1420464_s_at - -
          Mouse Genome 430A 2.0 Array 1424302_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424302_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424302_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6130022 - -
          Illumina mouse-6 v1.0 expression beadchip 6130022 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006130022 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1260379 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1260379 - -
          mouse-6 v1.1 (Illumina) scl018733.1_24-S - -
          Illumina Sentrix mouse-6.v1 bead array 6130022 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl018733.1_24-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6130022 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1260379 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0