Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01701417_s1
          See other HIST3H2A GE Assays ›
          Gene Symbol:
          Hist3h2a
          Gene Name:
          histone cluster 3, H2a
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 58954685 - 58955192 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01701417_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hist3h2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK051448.1 - 1844 102
          AK082083.1 - 1847 102

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27833727 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27833728 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27833729 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P615375 AK082083.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P615375 AK087537.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P615375 BC058119.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P615375 BC063781.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P615375 NM_178218.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P615375 AK082083.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P615375 AK087537.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P615375 BC058119.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P615375 BC063781.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P615375 NM_178218.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 244982 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P615375 AK082083.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P615375 AK087537.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P615375 BC058119.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P615375 BC063781.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P615375 NM_178218.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P615375 AK082083.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P615375 AK087537.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P615375 BC058119.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P615375 BC063781.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P615375 NM_178218.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P184352 AK082083.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P184352 AK087537.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P184352 BC058119.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P184352 BC063781.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P184352 NM_178218.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27833738 - -
          Whole Mouse Genome G4122A A_52_P184352 AK082083.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P184352 AK087537.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P184352 BC058119.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P184352 BC063781.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P184352 NM_178218.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615375 AK082083.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615375 AK087537.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615375 BC058119.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615375 BC063781.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615375 NM_178218.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615375 AK082083.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615375 AK087537.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615375 BC058119.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615375 BC063781.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P615375 NM_178218.3 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09483202 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09483210 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09483208 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09483214 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27833728 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27833727 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27833729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27833767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833729 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27833739 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10376456 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376447 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376457 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376458 - -
          Affymetrix HT MG-430 PM Array Plate 1455712_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1435865_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1435866_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10376455 - -
          Affymetrix Mouse Gene 1.1 ST Array 10376446 - -
          Mouse Genome 430 2.0 Array 1455712_PM_at - -
          Mouse Genome 430 2.0 Array 1440932_at - -
          Mouse Genome 430 2.0 Array 1435865_PM_at - -
          Mouse Genome 430 2.0 Array 1455712_at - -
          Mouse Genome 430 2.0 Array 1440932_PM_at - -
          Mouse Genome 430 2.0 Array 1435865_at - -
          Mouse Genome 430 2.0 Array 1435866_PM_s_at - -
          Mouse Genome 430 2.0 Array 1435866_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 319162_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 319162_PM_at - -
          Mouse Expression Set 430 1435865_PM_at - -
          Mouse Expression Set 430 1427507_at - -
          Mouse Expression Set 430 1435865_at - -
          Mouse Expression Set 430 1435866_s_at - -
          Mouse Expression Set 430 1435866_PM_s_at - -
          Mouse Expression Set 430 1427507_PM_at - -
          Mouse Expression Set 430 1455712_PM_at - -
          Mouse Expression Set 430 1440932_PM_at - -
          Mouse Expression Set 430 1440932_at - -
          Mouse Expression Set 430 1455712_at - -
          Murine Genome U74v2 Set 94805_f_at - -
          Murine Genome U74v2 Set 94805_PM_f_at - -
          Murine Genome U74v2 Set 167001_at - -
          Murine Genome U74v2 Set 167001_PM_at - -
          Mouse Genome 430A 2.0 Array 1435866_s_at - -
          Mouse Genome 430A 2.0 Array 1435865_at - -
          Mouse Genome 430A 2.0 Array 1427507_PM_at - -
          Mouse Genome 430A 2.0 Array 1435865_PM_at - -
          Mouse Genome 430A 2.0 Array 1435866_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1427507_at - -
          Affymetrix HT Mouse Genome 430A Array 1435866_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1435865_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435865_at - -
          Affymetrix HT Mouse Genome 430A Array 1435866_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1440932_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455712_at - -
          Affymetrix HT Mouse Genome 430B Array 1455712_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440932_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 653349 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105130064 - -
          Illumina mouse-6 v1.0 expression beadchip 105130064 - -
          Illumina mouse-6 v1.0 expression beadchip 1580504 - -
          Illumina mouse-6 v1.0 expression beadchip 106590338 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001580504 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2629289 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2629289 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2488159 - -
          mouse-6 v1.1 (Illumina) scl13126.1.1_324-S - -
          mouse-6 v1.1 (Illumina) scl00319162.1_289-S - -
          Illumina Sentrix mouse-6.v1 bead array 106590338 - -
          Illumina Sentrix mouse-6.v1 bead array 1580504 - -
          Illumina Sentrix mouse-6.v1 bead array 105130064 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00319162.1_289-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106590338 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1580504 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105130064 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2629289 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2629289 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0