Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01701525_m1
          See other SMIM5 GE Assays ›
          Gene Symbol:
          Smim5
          Gene Name:
          small integral membrane protein 5
          Gene Aliases:
          2210020M01Rik
          Chromosome Location:
          Chr.11: 115899512 - 115906269 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01701525_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smim5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183259.3 NP_899082.1 1 - 2 201 81
          GenBank mRNA AK004370.1 - 1 - 2 134 81
          AK020450.1 - 1 - 2 163 81
          BC032884.1 - 1 - 2 375 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001911.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK004370.1, AK020450.1, NM_183259.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smim5

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P295104 BC065414.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P295104 NM_183259.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P295104 AK004370.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P295104 AK020450.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P295104 AW124215.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P295104 BC065414.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P295104 NM_183259.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P295104 AK004370.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P295104 AK020450.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P295104 AW124215.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120628 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117009 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136821 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P295104 BC065414.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P295104 NM_183259.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P295104 AK004370.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P295104 AK020450.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P295104 AW124215.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P295104 BC065414.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P295104 NM_183259.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P295104 AK004370.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P295104 AK020450.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P295104 AW124215.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28156077 - -
          Whole Mouse Genome G4122A A_52_P295104 BC065414.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295104 NM_183259.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295104 AK004370.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295104 AK020450.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295104 AW124215.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295104 BC065414.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295104 NM_183259.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295104 AK004370.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295104 AK020450.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P295104 AW124215.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113721 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09398705 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09170930 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09398674 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09398665 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09669070 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10382697 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382694 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382693 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382696 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382695 - -
          Affymetrix HT MG-430 PM Array Plate 1435378_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10382692 - -
          Mouse Genome 430 2.0 Array 1435378_PM_at - -
          Mouse Genome 430 2.0 Array 1435378_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66528_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66528_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100018140.mm.1 TC1100001911.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100018140.mm.1 NM_183259.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018140.mm.1 NM_183259.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018135.mm.1 TC1100001911.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100018135.mm.1 NM_183259.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018135.mm.1 NM_183259.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018138.mm.1 TC1100001911.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100018138.mm.1 NM_183259.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018138.mm.1 NM_183259.3 Refseq
          Mouse Expression Set 430 1435378_PM_at - -
          Mouse Expression Set 430 1435378_at - -
          Affymetrix HT Mouse Genome 430B Array 1435378_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435378_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 461120 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4590273 - -
          Illumina mouse-6 v1.0 expression beadchip 4590273 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004590273 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2643326 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2643326 - -
          mouse-6 v1.1 (Illumina) scl40718.3_48-S - -
          Illumina Sentrix mouse-6.v1 bead array 4590273 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40718.3_48-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4590273 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2643326 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2643326 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0