Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01702692_m1
          See other GJA6 GE Assays ›
          Gene Symbol:
          Gja6
          Gene Name:
          gap junction protein, alpha 6
          Gene Aliases:
          A530015O12Rik, Cnx33, Cx33, Gja-6
          Chromosome Location:
          Chr.X: 160902116 - 160907052 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01702692_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gja6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001496.2 NP_001001496.1 1 - 2 144 112
          GenBank mRNA AK040693.1 - 1 - 2 104 112
          AK146838.1 - 1 - 2 144 112
          AK168953.1 - 1 - 2 144 112
          BC117760.2 - 1 - 2 87 40090837 112

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00003281.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK040693.1, AK146838.1, AK168953.1, BC117760.2, NM_001001496.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gja6

          Gene Ontology Categories:

          Function(s)

          cell communication

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P434059 BC117760.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P434059 AK168953.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P434059 NM_001001496.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P434059 AK040693.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P434059 AK146838.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P434059 BC117760.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P434059 AK168953.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P434059 NM_001001496.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P434059 AK040693.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P434059 AK146838.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100160 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P434059 BC117760.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P434059 AK168953.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P434059 NM_001001496.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P434059 AK040693.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P434059 AK146838.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P434059 BC117760.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P434059 AK168953.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P434059 NM_001001496.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P434059 AK040693.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P434059 AK146838.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434059 BC117760.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434059 AK168953.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434059 NM_001001496.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434059 AK040693.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434059 AK146838.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434059 BC117760.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434059 AK168953.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434059 NM_001001496.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434059 AK040693.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P434059 AK146838.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32782148 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100160 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09476419 - -
          Whole Mouse Genome G4122A A_51_P434059 BC117760.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434059 AK168953.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434059 NM_001001496.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434059 AK040693.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434059 AK146838.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434059 BC117760.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434059 AK168953.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434059 NM_001001496.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434059 AK040693.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P434059 AK146838.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103819 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100160 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14637848 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14637846 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14660552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32782157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15172130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15092753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14994846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14981029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14838217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14981015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32782168 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10607642 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607641 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607640 - -
          Affymetrix Mouse Gene 1.1 ST Array 10607639 - -
          Affymetrix Mouse Clariom S Array PSR0X00020524.mm.1 TC0X00003281.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00020524.mm.1 NM_001001496.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00020524.mm.1 NM_001001496.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00020525.mm.1 TC0X00003281.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00020525.mm.1 NM_001001496.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00020525.mm.1 NM_001001496.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00020522.mm.1 TC0X00003281.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00020522.mm.1 NM_001001496.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00020522.mm.1 NM_001001496.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00020523.mm.1 TC0X00003281.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00020523.mm.1 NM_001001496.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00020523.mm.1 NM_001001496.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 785058 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 106550167 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2561376 - -
          Illumina Sentrix mouse-6.v1 bead array 106550167 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106550167 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2946541 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2946541 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0