Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01718316_m1
          See other 4933405D12RIK GE Assays ›
          Gene Symbol:
          4933405D12Rik
          Gene Name:
          RIKEN cDNA 4933405D12 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 122882592 - 122924116 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01718316_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933405D12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046036.1 4 - 5 502 67
          GenBank mRNA AK016661.1 - 504 67
          AK045097.1 - 4 - 5 503 67
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB294941.2 - 3 - 4 440 Chr. 3 - 122882592 - 122919780 [-] on Build GRCm38 67
          GenBankmRNA BY716349.1 - 4 - 5 504 Chr. 3 - 122865158 - 122924116 [-] on Build GRCm38 67
          GenBankmRNA BB643330.1 - 4 - 5 500 Chr. 3 - 122889267 - 122924114 [-] on Build GRCm38 67

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream phosphodiesterase 5A,
          cGMP-specific
          Pde5a Cgbpde,
          Cn5n,
          Pde5,
          Pde5a1
          -29.9kb Anti-sense Chr. 3 - 122729158 - 122859374 on NCBI Build GRCm38
          Downstream fatty acid binding protein 2,
          intestinal
          Fabp2 Fabpi,
          I-FABP
          4.8kb Anti-sense Chr. 3 - 122895072 - 122899506 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002866.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2350553 AK016661.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707719 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 803890 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925102 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 881278 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 153121 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 207144 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 325830 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73576 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 423881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 410558 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 829807 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 856423 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 171779 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444281 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 225564 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 489390 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 122066 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 886762 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 305246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529696 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 417150 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 257964 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 329804 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351201 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532062 AK012158.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532062 AK078091.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532062 AK088426.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532062 BC014810.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532062 BC029230.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532062 NM_028151.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669800 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 173945 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 154331 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 38808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126113 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 65382 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158734 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05055937 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117364 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155131 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05055536 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05055560 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00743961 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00744020 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158734 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05055514 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05055995 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05055955 - -
          Whole Mouse Genome G4122A A_52_P242609 AK016661.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101784 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155131 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05055514 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158734 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117364 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431736_PM_at - -
          Mouse Genome 430 2.0 Array 1431736_PM_at - -
          Mouse Genome 430 2.0 Array 1431736_at - -
          Affymetrix Mouse Clariom S Array PSR0300022670.mm.1 TC0300002866.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300022670.mm.1 NR_046036.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022670.mm.1 NR_046036.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022671.mm.1 TC0300002866.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300022671.mm.1 NR_046036.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300022671.mm.1 NR_046036.1 Refseq
          Mouse Expression Set 430 1431736_at - -
          Mouse Expression Set 430 1431736_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431736_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431736_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 682553 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0