Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01718413_m1
          See other A330023F24RIK GE Assays ›
          Gene Symbol:
          A330023F24Rik
          Gene Name:
          RIKEN cDNA A330023F24 gene
          Gene Aliases:
          C030002C11Rik
          Chromosome Location:
          Chr.1: 195017399 - 195037908 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01718413_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A330023F24Rik
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015566.2
          7 - 8 1102 79
          GenBank mRNA AK045637.1 -
          7 - 8 1103 79
          BC058715.1 - LIT3442
          C030002C11Rik
          2 - 3 343 79
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CB723210.1 - 2 - 3 344 Chr. 1 - 195024109 - 195034161 [+] on Build GRCm38 79

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          16897
          Gm16897 C030002c11rik -54.9kb Anti-sense Chr. 1 - 194955759 - 194976959 on NCBI Build GRCm38
          Downstream microRNA 29b-2 Mir29b-2 Mirn29b-2,
          mir-29b-2
          111.0 Sense Chr. 1 - 195037040 - 195037120 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001883.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2192397 AK171382.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 870002 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132538 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358611 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783398 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 760796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 781031 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 163988 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561224 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 849842 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841148 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 288124 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 205851 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 584790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 128536 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 341390 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 1669 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 145797 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 136706 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 488726 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 443660 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 93915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928617 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 861620 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 146634 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 1410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 197152 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 869251 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 503469 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 906412 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 163907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 337642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 469769 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427174 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787834 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 947014 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481546 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765037 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64608 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167394 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 43673 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166252 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102414 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167486 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04409547 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04409518 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00290599 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04409463 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00290546 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04409568 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04409581 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00290573 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167486 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04409537 - -
          Whole Mouse Genome G4122A A_52_P614832 NR_015566.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P614832 AK045637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P614832 BC058715.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132458 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00290546 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167486 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04409581 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00290573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20959125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20959115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20959145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20959109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00453497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20959076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00070946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00068823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20959119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00812901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00070947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00812902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20959127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20959128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00068847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20959130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00068819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20959118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00948349 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20959142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00523474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00068843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00068851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00068846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00068866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20959135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00749827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20959069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00812898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00068859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00224547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00068834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20959108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00453499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20959113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00201133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20959110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00201114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20959067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00201135 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10352919 - -
          Affymetrix Mouse Gene 1.0 ST Array 10352917 - -
          Affymetrix Mouse Gene 1.0 ST Array 10352915 - -
          Affymetrix HT MG-430 PM Array Plate 1460033_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10352918 - -
          Affymetrix Mouse Gene 1.1 ST Array 10352914 - -
          Affymetrix Mouse Gene 1.1 ST Array 10352916 - -
          Mouse Genome 430 2.0 Array 1460033_at - -
          Mouse Genome 430 2.0 Array 1460033_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100015674.mm.1 TC0100001883.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100015674.mm.1 NR_015566.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015674.mm.1 NR_015566.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015674.mm.1 NR_029745.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015674.mm.1 NR_029745.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015675.mm.1 TC0100001883.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100015675.mm.1 NR_015566.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015675.mm.1 NR_015566.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015669.mm.1 TC0100001883.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100015669.mm.1 NR_015566.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015669.mm.1 NR_015566.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015670.mm.1 TC0100001883.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100015670.mm.1 NR_015566.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015670.mm.1 NR_015566.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015671.mm.1 TC0100001883.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100015671.mm.1 NR_015566.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015671.mm.1 NR_015566.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015671.mm.1 NR_029809.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015671.mm.1 NR_029809.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015673.mm.1 TC0100001883.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100015673.mm.1 NR_015566.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015673.mm.1 NR_015566.2 Refseq
          Mouse Expression Set 430 1460033_PM_at - -
          Mouse Expression Set 430 1460033_at - -
          Affymetrix HT Mouse Genome 430B Array 1460033_at - -
          Affymetrix HT Mouse Genome 430B Array 1460033_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 459042 - -
          ABI Mouse Genome Survey Microarray 718966 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6200390 - -
          MouseWG-6 v2 Expression BeadChip 2480471 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0