Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01718423_s1
          See other CAMK2N1 GE Assays ›
          Gene Symbol:
          Camk2n1
          Gene Name:
          calcium/calmodulin-dependent protein kinase II inhibitor 1
          Gene Aliases:
          1810006K23Rik, AI848599, AI891706
          Chromosome Location:
          Chr.4: 138455148 - 138460126 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01718423_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Camk2n1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025451.2 NP_079727.1 2 1581 101
          GenBank mRNA AK007352.1 - 1 831 101
          AY523601.1 - 2 1543 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001728.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein kinase activity
          protein kinase inhibitor activity
          calcium-dependent protein kinase inhibitor activity
          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345849 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345844 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345845 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345843 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345840 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345841 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345842 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345847 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345846 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345839 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64687 NM_025451.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64687 AK007352.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64687 AK021100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64687 AY523601.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64687 NM_025451.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64687 AK007352.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64687 AK021100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P64687 AY523601.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 376806 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960242 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 591819 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 30698 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160578 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64687 NM_025451.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64687 AK007352.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64687 AK021100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64687 AY523601.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64687 NM_025451.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64687 AK007352.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64687 AK021100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P64687 AY523601.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160578 - -
          Whole Mouse Genome G4122A A_52_P64687 NM_025451.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P64687 AK007352.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P64687 AK021100.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P64687 AY523601.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P64687 NM_025451.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P64687 AK007352.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P64687 AK021100.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P64687 AY523601.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100713 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160578 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345844 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345842 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345846 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345849 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345847 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345843 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345845 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345840 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345841 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23345839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345907 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03335153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03410647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03579155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03893660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03594664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345890 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03608786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03701026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03410644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03815902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03608796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03570892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03410648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03595727 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03608784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03570891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03788970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03531675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04025092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03579153 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03608794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03844859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03893659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03630721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03701025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03579141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345895 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345902 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10509569 - -
          Affymetrix Mouse Gene 1.0 ST Array 10509570 - -
          Affymetrix HT MG-430 PM Array Plate 1440455_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456609_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10509568 - -
          Mouse Genome 430 2.0 Array 1456609_PM_at - -
          Mouse Genome 430 2.0 Array 1456609_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66259_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66259_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400014124.mm.1 TC0400001728.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014124.mm.1 NM_025451.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014124.mm.1 NM_025451.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014126.mm.1 TC0400001728.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014126.mm.1 NM_025451.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014126.mm.1 NM_025451.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014127.mm.1 TC0400001728.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014127.mm.1 NM_025451.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014127.mm.1 NM_025451.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014125.mm.1 TC0400001728.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400014125.mm.1 NM_025451.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400014125.mm.1 NM_025451.2 Refseq
          Mouse Expression Set 430 1456609_at - -
          Mouse Expression Set 430 1456609_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456609_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440455_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456609_at - -
          Affymetrix HT Mouse Genome 430B Array 1440455_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 466173 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2487046 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2810963 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2810963 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0