Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01719003_m1
          See other 6030440G07RIK GE Assays ›
          Gene Symbol:
          6030440G07Rik
          Gene Name:
          RIKEN cDNA 6030440G07 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 110994398 - 111013872 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01719003_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 6030440G07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_036598.1 1 - 2 310 80
          GenBank mRNA AK020059.1 - 1 - 2 313 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001223.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998219 AK020059.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 730881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 65242 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647271 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 115491 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427178 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648789 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 379270 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 154079 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 141228 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102415 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108064 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998219 AK020059.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07750462 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108064 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07750481 - -
          Whole Mouse Genome G4122A A_52_P850915 AK020059.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134345 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108064 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431495_PM_at - -
          Mouse Genome 430 2.0 Array 1431495_at - -
          Mouse Genome 430 2.0 Array 1431495_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77878_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77878_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200008637.mm.1 TC1200001223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008637.mm.1 NR_036598.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008637.mm.1 NR_036598.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008638.mm.1 TC1200001223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008638.mm.1 NR_036598.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008638.mm.1 NR_036598.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008639.mm.1 TC1200001223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008639.mm.1 NR_036598.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008639.mm.1 NR_036598.1 Refseq
          Mouse Expression Set 430 1431495_PM_at - -
          Mouse Expression Set 430 1431495_at - -
          Murine Genome U74v2 Set 168370_PM_at - -
          Murine Genome U74v2 Set 168370_at - -
          Affymetrix HT Mouse Genome 430B Array 1431495_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431495_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 607115 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0