Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01719348_m1
          See other GM5427 GE Assays ›
          Gene Symbol:
          Gm5427
          Gene Name:
          predicted gene 5427
          Gene Aliases:
          EG432494
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01719348_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm5427
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK076335.1 - 100 71
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB612635.1 - 1 - 2 100 Chr. 10 - 98655706 - 98699710 [-] on Build GRCm38 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          34297
          Gm34297 -88.2kb Anti-sense Chr. 10 - 98537488 - 98567511 on NCBI Build GRCm38
          Downstream ATPase,
          Ca++ transporting,
          plasma membrane 1
          Atp2b1 2810442I22Rik,
          E130111D10Rik,
          Pmca1
          214.6kb Anti-sense Chr. 10 - 98914359 - 99026143 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P123835 AK076335.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426217 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 191751 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292675 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530608 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 338032 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 876025 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 614096 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 259749 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 705260 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 691268 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495331 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 830854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 280155 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 145625 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 71587 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 69093 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 62286 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P123835 AK076335.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07165611 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07165588 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07165656 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02199256 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123835 AK076335.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07165656 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02199256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382539 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08844504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382397 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08767952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08767939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08458418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08767936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27382400 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 613682 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0