Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01721930_m1
          See other GM29676 GE Assays ›
          Gene Symbol:
          Gm29676
          Gene Name:
          predicted gene, 29676
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 44250092 - 44260123 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01721930_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm29676
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK035008.1 - 115 98
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB627242.1 - 1 - 2 115 Chr. 13 - 44250524 - 44252670 [-] on Build GRCm38 98

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA A330076C08 gene A330076C08Rik -34.0kb Sense Chr. 13 - 44193589 - 44216487 on NCBI Build GRCm38
          Downstream RIKEN cDNA 1700029N11 gene 1700029N11Rik 187.1kb Anti-sense Chr. 13 - 44439727 - 44457567 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P123028 AK035008.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P123028 AK035008.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123028 AK035008.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P28995031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28994990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10958960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28994984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28995041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28994982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28995038 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11070057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28994983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10826831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28994987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28995037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28995040 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28994995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11042364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28995030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28995028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10639518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28995050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28994993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28995046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11070055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28995035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28995049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28995026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10527368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28994992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28994996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28994998 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28995045 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 496663 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0