Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01722271_g1
          See other TSR3 GE Assays ›
          Gene Symbol:
          Tsr3
          Gene Name:
          TSR3 20S rRNA accumulation
          Gene Aliases:
          0610007P22Rik, 1110014B11Rik, AL022759
          Chromosome Location:
          Chr.17: 25240170 - 25242799 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01722271_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsr3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163718.1 NP_001157190.1 5 - 6 823 78
          NM_026676.3 NP_080952.2 5 - 6 844 78
          GenBank mRNA AK158743.1 - 5 - 6 845 78
          AK169072.1 - 4 - 5 1179 78
          AK170074.1 - 5 - 6 813 78
          BC006899.1 - 2 - 3 189 78
          BC027751.1 - 5 - 6 914 5011894 78
          BC089019.1 - 5 - 6 793 30551363 78
          CA979987.1 - 5 - 6 772 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000367.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tsr3

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA processing
          biological_process
          maturation of SSU-rRNA
          ribosome biogenesis
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31102486 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31102487 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31102484 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148833 AK169072.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148833 BC027751.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148833 BC038884.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148833 AK003675.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139901 AK122358.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139901 NM_001163270.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139901 BC022659.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139901 AK169072.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139901 AK170074.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 27011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 6374 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137081 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139901 AK122358.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139901 NM_001163270.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139901 BC022659.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139901 AK169072.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139901 AK170074.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102489 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102508 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102528 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102488 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102518 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102486 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102516 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102496 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102487 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102522 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102498 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31102494 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137081 - -
          Whole Mouse Genome G4122A A_52_P552893 AK169072.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P552893 AK170074.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P552893 BC006899.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P552893 BC027751.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P552893 BC038884.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113013 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137081 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13506773 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13506770 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31102484 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31102487 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31102486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31102487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13561714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13506752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13346049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13346007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13561721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13346004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13346013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102522 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13153519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102487 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102515 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13561716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102566 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13346010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13346018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31102497 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442755 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442756 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442754 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442758 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442757 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442760 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581958 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442753 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442761 - -
          Affymetrix HT MG-430 PM Array Plate 1431132_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1437969_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10581957 - -
          Affymetrix Mouse Gene 1.1 ST Array 10442752 - -
          Mouse Genome 430 2.0 Array 1437969_PM_s_at - -
          Mouse Genome 430 2.0 Array 1431132_x_at - -
          Mouse Genome 430 2.0 Array 1437969_s_at - -
          Mouse Genome 430 2.0 Array 1431132_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68327_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68327_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700003052.mm.1 TC1700000367.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003052.mm.1 NM_001163718.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003052.mm.1 NM_001163718.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003052.mm.1 NM_026676.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003052.mm.1 NM_026676.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003054.mm.1 TC1700000367.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700003054.mm.1 NM_001163718.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003054.mm.1 NM_001163718.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003054.mm.1 NM_026676.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700003054.mm.1 NM_026676.3 Refseq
          Mouse Expression Set 430 1437969_PM_s_at - -
          Mouse Expression Set 430 1437969_s_at - -
          Murine Genome U74v2 Set 103372_at - -
          Murine Genome U74v2 Set 103372_PM_at - -
          Mouse Genome 430A 2.0 Array 1437969_s_at - -
          Mouse Genome 430A 2.0 Array 1437969_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1437969_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1431132_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1437969_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1431132_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 130193 - -
          MouseWG-6 v2 Expression BeadChip 106650369 - -
          Illumina mouse-6 v1.0 expression beadchip 3800433 - -
          Illumina mouse-6 v1.0 expression beadchip 130193 - -
          Illumina mouse-6 v1.0 expression beadchip 6020750 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003800433 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000130193 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006020750 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2634566 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215157 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1216623 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216623 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2634566 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215157 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252234 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2480021 - -
          mouse-6 v1.1 (Illumina) scl000098.1_12-S - -
          mouse-6 v1.1 (Illumina) scl50988.5.36_204-S - -
          Illumina Sentrix mouse-6.v1 bead array 3800433 - -
          Illumina Sentrix mouse-6.v1 bead array 130193 - -
          Illumina Sentrix mouse-6.v1 bead array 6020750 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50988.5.36_204-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000098.1_12-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 130193 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6020750 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3800433 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2634564 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1216623 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1215157 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2634564 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1216623 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215157 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000098.1_12-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50988.5.36_204-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0