Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01961603_s1
          See other AA415398 GE Assays ›
          Gene Symbol:
          AA415398
          Gene Name:
          expressed sequence AA415398
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 119530308 - 119538769 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01961603_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AA415398
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004178.1 NP_001004178.1 4 1043 108
          GenBank mRNA BC064115.1 - 1 309 3417656 108
          BC076608.1 - 4 1043 30625085 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003430.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene AA415398

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23246057 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23246054 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23246055 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23246056 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126842 BC076608.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126842 NM_001004178.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126842 BC064115.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240506 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649436 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 2619 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 419683 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 706291 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 34365 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126842 BC076608.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126842 NM_001004178.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126842 BC064115.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23246051 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00971342 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05376741 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126842 BC076608.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126842 NM_001004178.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126842 BC064115.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128831 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05376741 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03626057 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03626051 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03626059 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03626060 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03626052 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23246054 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23246055 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23246056 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23246057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23245963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23245993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23245961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23245989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23245986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23245975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23245966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23245960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23245959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23245970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23245973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23245958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23246031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23245976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23245965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23245953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23245979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03626060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03623930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23245975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03626066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03634658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23245952 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03344764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23245959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23245970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246021 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23245966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03685590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23245968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03252271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03730893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23245976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246038 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03344766 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23245961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246017 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23245960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23246044 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10515931 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515934 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515932 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515933 - -
          Affymetrix HT MG-430 PM Array Plate 1439499_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10515930 - -
          Mouse Genome 430 2.0 Array 1439499_at - -
          Mouse Genome 430 2.0 Array 1439499_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 433752_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 433752_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400027862.mm.1 TC0400003430.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027862.mm.1 NM_001004178.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027862.mm.1 NM_001004178.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027866.mm.1 TC0400003430.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027866.mm.1 NM_001004178.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027866.mm.1 NM_001004178.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027861.mm.1 TC0400003430.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027861.mm.1 NM_001004178.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027861.mm.1 NM_001004178.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027868.mm.1 TC0400003430.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027868.mm.1 NM_001004178.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027868.mm.1 NM_001004178.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027858.mm.1 TC0400003430.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027858.mm.1 NM_001004178.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027858.mm.1 NM_001004178.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027860.mm.1 TC0400003430.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027860.mm.1 NM_001004178.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027860.mm.1 NM_001004178.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027864.mm.1 TC0400003430.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027864.mm.1 NM_001004178.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027864.mm.1 NM_001004178.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027859.mm.1 TC0400003430.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027859.mm.1 NM_001004178.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027859.mm.1 NM_001004178.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027863.mm.1 TC0400003430.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027863.mm.1 NM_001004178.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027863.mm.1 NM_001004178.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027869.mm.1 TC0400003430.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400027869.mm.1 NM_001004178.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400027869.mm.1 NM_001004178.1 Refseq
          Mouse Expression Set 430 1439499_PM_at - -
          Mouse Expression Set 430 1439499_at - -
          Affymetrix HT Mouse Genome 430B Array 1439499_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439499_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160592 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160592 - -
          mouse-6 v1.1 (Illumina) GI_51921350-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160592 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160592 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) GI_51921350-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0