Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02021559_s1
          See other B130011K05RIK GE Assays ›
          Gene Symbol:
          B130011K05Rik
          Gene Name:
          RIKEN cDNA B130011K05 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02021559_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B130011K05Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK044916.1 - 2 3205 149

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream peripheral myelin protein 22 Pmp22 22kDa,
          Gas-3,
          HNPP,
          Tr,
          trembler
          -256.4kb Sense Chr. 11 - 63128982 - 63159547 on NCBI Build GRCm38
          Downstream heparan sulfate (glucosamine) 3-O-sulfotransferase 3B1 Hs3st3b1 3-OST-3B,
          3OST3B1,
          3Ost3b,
          AU041882,
          AW536289,
          Hs3st3b,
          m3-OST-3B
          466.8kb Anti-sense Chr. 11 - 63884693 - 63922284 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 306787 - -
          Whole Mouse Genome G4122A A_52_P923314 AK044916.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08983540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27859465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27859473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27859463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27859455 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27859477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27859461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27859456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27859467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27859474 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27859460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09175809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27859470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08983538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09175817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27859454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27859453 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27859464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27859458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08983535 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 470538 - -
          ABI Mouse Genome Survey Microarray 761464 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100540593 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0