Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02024950_s1
          See other 9330185C12RIK GE Assays ›
          Gene Symbol:
          9330185C12Rik
          Gene Name:
          RIKEN cDNA 9330185C12 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02024950_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9330185C12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK020392.1 - 3 724 141

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream dermatan sulfate epimerase-like Dsel 9330132E09Rik,
          DS-epi2
          -2.0mb Sense Chr. 1 - 111858701 - 111864918 on NCBI Build GRCm38
          Downstream contactin associated protein-like 5A Cntnap5a Caspr5-1,
          EG636808
          1.8mb Anti-sense Chr. 1 - 115684497 - 116587323 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 661227 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 535557 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 159506 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 917401 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 418725 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 393109 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127759 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 151660 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 470242 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 603670 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453797 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 315279 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 405662 AK007286.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490370 NM_134177.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 679656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764721 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728446 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 417364 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16663 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12842 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79411 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04228092 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04228188 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04228125 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00160644 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00160625 - -
          Whole Mouse Genome G4122A A_52_P681722 AK020392.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123707 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04228125 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00160644 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1454220_PM_at - -
          Mouse Genome 430 2.0 Array 1454220_at - -
          Mouse Genome 430 2.0 Array 1454220_PM_at - -
          Mouse Expression Set 430 1454220_at - -
          Mouse Expression Set 430 1454220_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454220_at - -
          Affymetrix HT Mouse Genome 430B Array 1454220_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 793850 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0