Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900015426.mm.1 |
TC1900001745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900015426.mm.1 |
NM_030197.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015426.mm.1 |
NM_030197.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015428.mm.1 |
TC1900001745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900015428.mm.1 |
NM_001172096.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015428.mm.1 |
NM_001172096.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015428.mm.1 |
NM_001172097.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015428.mm.1 |
NM_001172097.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015428.mm.1 |
NM_030197.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015428.mm.1 |
NM_030197.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015429.mm.1 |
TC1900001745.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900015429.mm.1 |
NM_001172096.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015429.mm.1 |
NM_001172096.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015429.mm.1 |
NM_001172097.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015429.mm.1 |
NM_001172097.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015429.mm.1 |
NM_030197.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015429.mm.1 |
NM_030197.2 |
Refseq |