Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02393486_mH
          See other GM10319 GE Assays ›
          Gene Symbol:
          Gm10319
          Gene Name:
          murinoglobulin pseudogene
          Gene Aliases:
          EG381806, ENSMUSG00000071204
          Chromosome Location:
          Chr.6: 122136628 - 122150958 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02393486_mH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm10319
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003624.2 2 - 3 420 152
          GenBank mRNA BC061166.1 - 2 - 3 420 152

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001375.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105416 NR_003624.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2105416 BC061166.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2116485 NR_003624.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2116485 BC061166.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 76871 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42993 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105416 NR_003624.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2105416 BC061166.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2116485 NR_003624.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2116485 BC061166.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P202367 NR_003624.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P202367 BC061166.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P24807229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24807249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24807259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05609360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05243208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24807210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05243222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24807213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24807235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24807277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24807233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05243213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05243220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05803569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05243202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24807219 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10541454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541443 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541442 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541445 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541444 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541449 - -
          Affymetrix Mouse Gene 1.1 ST Array 10541441 - -
          Affymetrix Mouse Clariom S Array PSR0600011083.mm.1 TC0600001375.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011083.mm.1 NR_003624.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011077.mm.1 TC0600001375.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011077.mm.1 NR_003624.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011078.mm.1 TC0600001375.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011078.mm.1 NR_003624.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011080.mm.1 TC0600001375.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011080.mm.1 NR_003624.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011081.mm.1 TC0600001375.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011081.mm.1 NR_003624.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011082.mm.1 TC0600001375.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011082.mm.1 NR_003624.2 Refseq
          Murine Genome U74v2 Set 135778_PM_at - -
          Murine Genome U74v2 Set 135778_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161901 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161901 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161901 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0