Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02528717_g1
          See other B230354K17RIK GE Assays ›
          Gene Symbol:
          B230354K17Rik
          Gene Name:
          RIKEN cDNA B230354K17 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 45433736 - 45492165 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02528717_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B230354K17Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK083460.1 - 2 1158 77
          AK140548.1 - 2 1144 77
          AK171144.1 - 2 1158 77
          BC027345.1 - 1 361 77

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P101238 BB440272.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P101238 AK083460.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P101238 BB440272.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P101238 AK083460.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31201122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31201138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31201124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31201136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31201133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31201130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31201132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31201128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31201125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31201126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31201127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31201135 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31201129 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08787114 - -
          Whole Mouse Genome G4122A A_52_P101238 BB440272.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P101238 AK083460.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P101238 BB440272.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P101238 AK083460.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P838078 AK171144.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P838078 AK140548.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P838078 AK162357.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P838078 AK045689.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P838078 AK046225.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110400 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445375 - -
          Affymetrix HT MG-430 PM Array Plate 1424515_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1424516_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10445373 - -
          Mouse Genome 430 2.0 Array 1424515_at - -
          Mouse Genome 430 2.0 Array 1424516_at - -
          Mouse Genome 430 2.0 Array 1424516_PM_at - -
          Mouse Genome 430 2.0 Array 1424515_PM_at - -
          Mouse Expression Set 430 1424515_at - -
          Mouse Expression Set 430 1424515_PM_at - -
          Mouse Expression Set 430 1424516_at - -
          Mouse Expression Set 430 1424516_PM_at - -
          Mouse Genome 430A 2.0 Array 1424515_at - -
          Mouse Genome 430A 2.0 Array 1424516_PM_at - -
          Mouse Genome 430A 2.0 Array 1424516_at - -
          Mouse Genome 430A 2.0 Array 1424515_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424516_at - -
          Affymetrix HT Mouse Genome 430A Array 1424516_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424515_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424515_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 353516 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 450746 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000450746 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2594256 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2594256 - -
          mouse-6 v1.1 (Illumina) scl50691.4_417-S - -
          Illumina Sentrix mouse-6.v1 bead array 450746 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50691.4_417-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 450746 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2594256 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0