Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02600697_s1
          See other ITPRIPL2 GE Assays ›
          Gene Symbol:
          Itpripl2
          Gene Name:
          inositol 1,4,5-triphosphate receptor interacting protein-like 2
          Gene Aliases:
          C130081G24, E030018N11Rik
          Chromosome Location:
          Chr.7: 118485111 - 118491975 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02600697_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Itpripl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033380.3 NP_001028552.1 1 5355 119
          GenBank mRNA AK086993.1 - 1 2826 119
          AK134593.1 - 1 5303 119
          AK137678.1 - 1 5357 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004172.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Itpripl2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458537 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458549 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458531 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458535 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458548 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458539 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458538 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458546 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458541 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458540 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458532 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458536 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458543 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458547 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458545 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458534 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458530 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458529 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458544 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458550 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046348 NM_001033380.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046348 AA182221.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060284 AK164927.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060284 AK081844.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060284 NM_001033380.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58431 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 844204 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490719 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118286 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060284 AK164927.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060284 AK081844.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060284 NM_001033380.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046348 NM_001033380.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046348 AA182221.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06339824 - -
          Whole Mouse Genome G4122A A_52_P110541 AK036744.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110541 AK086993.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110541 AK134593.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110541 AK137678.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P110541 NM_001033380.3 mouse_transcriptome
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458544 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458532 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458530 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458547 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458531 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458539 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458536 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458537 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458535 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458549 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458545 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458546 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458543 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458550 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458541 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458540 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458548 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458538 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458529 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25458534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25458454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458437 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458472 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458544 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06031431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06378673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06189784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06015786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06594274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06628365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06628366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06594275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458518 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06398122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06210087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06428373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06085036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06594278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458500 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06398114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458566 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25458465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05946572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06189758 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10567300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567302 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567301 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567298 - -
          Affymetrix HT MG-430 PM Array Plate 1435777_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10567299 - -
          Affymetrix Mouse Gene 1.1 ST Array 10567297 - -
          Mouse Genome 430 2.0 Array 1435777_PM_at - -
          Mouse Genome 430 2.0 Array 1435777_at - -
          Affymetrix Mouse Clariom S Array PSR0700034675.mm.1 TC0700004172.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700034675.mm.1 NM_001033380.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034675.mm.1 NM_001033380.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034676.mm.1 TC0700004172.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700034676.mm.1 NM_001033380.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700034676.mm.1 NM_001033380.3 Refseq
          Mouse Expression Set 430 1435777_PM_at - -
          Mouse Expression Set 430 1435777_at - -
          Affymetrix HT Mouse Genome 430B Array 1435777_at - -
          Affymetrix HT Mouse Genome 430B Array 1435777_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252004 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix