Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02621109_s1
          See other LPAR5 GE Assays ›
          Gene Symbol:
          Lpar5
          Gene Name:
          lysophosphatidic acid receptor 5
          Gene Aliases:
          GPR93, Gm1072, Gpr92, LPA5
          Chromosome Location:
          Chr.6: 125067842 - 125086129 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02621109_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lpar5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163268.1 NP_001156740.1 2 455 67
          NM_001163269.1 NP_001156741.1 2 390 67
          GenBank mRNA AK131863.1 - 2 391 67
          AY255621.1 - 1 68 67
          BC116702.1 - 2 355 40104865 67
          BC117528.1 - 2 355 40104864 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001443.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G protein-coupled receptor signaling pathway
          G protein-coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818553 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818549 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818547 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818551 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818548 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818550 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017699 BC116702.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017699 BC117528.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017699 NM_001163268.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017699 NM_001163269.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 223542 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 966524 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768183 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 33495 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 135776 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 624028 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 30642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 25582 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115045 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017699 BC116702.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017699 BC117528.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017699 NM_001163268.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017699 NM_001163269.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01438216 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115045 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01438209 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06057148 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115045 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818553 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818547 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818551 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818549 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818550 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24818548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05549048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818520 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818512 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24818518 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10541801 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541800 - -
          Affymetrix Mouse Gene 1.0 ST Array 10541802 - -
          Affymetrix Mouse Gene 1.1 ST Array 10541799 - -
          Affymetrix Mouse Clariom S Array PSR0600011738.mm.1 TC0600001443.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600011738.mm.1 NM_001163268.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011738.mm.1 NM_001163268.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011738.mm.1 NM_001163269.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600011738.mm.1 NM_001163269.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 774506 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2539669 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix