Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02764754_g1
          See other 2610005L07RIK GE Assays ›
          Gene Symbol:
          2610005L07Rik 6820431F20Rik
          Gene Name:
          cadherin 11 pseudogene
          cadherin 11 pseudogene
          Gene Aliases:
          2810038F24Rik, 6720476A01Rik
          -
          Chromosome Location:
          Chr.8: 20385782 - 20424814 on Build GRCm38
          Chromosome Location:
          Chr.8: 20268286 - 20297432 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm02764754_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2610005L07Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028428.1 5 1231 92
          GenBank mRNA AK009182.1 - 5 1050 92
          AK011323.1 - 5 1127 92
          AK017816.1 - 6 1127 92
          AK035547.1 - 2 339 92
          AK081322.1 - 5 1127 92
          AK131627.1 - 5 1232 92
          AK160987.1 - 1059 92
          BC086760.1 - 947 92
          BC086761.1 - 927 92
          Gene Symbol: 6820431F20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_030708.1 5 941 92
          GenBank mRNA AK135487.1 - 5 942 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800003240.mm.1, TC0800003241.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148001 AK135487.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148001 NR_030708.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148001 BC007164.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148001 AK081322.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148001 AK035547.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006930 BC025151.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006930 U80890.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148002 AK135487.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148002 NR_030708.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148002 BC086760.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148002 BC086761.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148002 AK160987.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2148001 AK135487.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2148001 NR_030708.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2148001 BC007164.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2148001 AK081322.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2148001 AK035547.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2148002 AK135487.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2148002 NR_030708.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2148002 BC086760.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2148002 BC086761.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2148002 AK160987.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006930 BC025151.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006930 U80890.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01689071 - -
          Whole Mouse Genome G4122A A_52_P341708 AK007507.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P640152 - -
          Whole Mouse Genome G4122A A_52_P335089 AK135487.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P335089 NR_030708.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P335089 BC025151.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P335089 AK160987.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P335089 AK081322.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P425634 - -
          Whole Mouse Genome G4122A A_52_P7105 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01689071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25704671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25704582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25704588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25704584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25704583 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10577411 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577415 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577403 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577407 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577417 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577396 - -
          Affymetrix Mouse Gene 1.0 ST Array 10570640 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577406 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577409 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577414 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577397 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577418 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577408 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577410 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577398 - -
          Affymetrix Mouse Gene 1.0 ST Array 10577416 - -
          Affymetrix HT MG-430 PM Array Plate 1457673_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440387_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1437717_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1429040_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1436740_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1452997_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429882_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429041_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10457223 - -
          Affymetrix Mouse Gene 1.1 ST Array 10570639 - -
          Affymetrix Mouse Gene 1.1 ST Array 10577412 - -
          Affymetrix Mouse Gene 1.1 ST Array 10577395 - -
          Affymetrix Mouse Gene 1.1 ST Array 10457223 - -
          Affymetrix Mouse Gene 1.1 ST Array 10577395 - -
          Affymetrix Mouse Gene 1.1 ST Array 10577412 - -
          Mouse Genome 430 2.0 Array 1429882_PM_at - -
          Mouse Genome 430 2.0 Array 1435470_PM_at - -
          Mouse Genome 430 2.0 Array 1457673_at - -
          Mouse Genome 430 2.0 Array 1429882_at - -
          Mouse Genome 430 2.0 Array 1436740_at - -
          Mouse Genome 430 2.0 Array 1440387_at - -
          Mouse Genome 430 2.0 Array 1429041_at - -
          Mouse Genome 430 2.0 Array 1440387_PM_at - -
          Mouse Genome 430 2.0 Array 1429040_at - -
          Mouse Genome 430 2.0 Array 1435470_at - -
          Mouse Genome 430 2.0 Array 1429041_PM_at - -
          Mouse Genome 430 2.0 Array 1437717_x_at - -
          Mouse Genome 430 2.0 Array 1452997_PM_at - -
          Mouse Genome 430 2.0 Array 1436740_PM_at - -
          Mouse Genome 430 2.0 Array 1452997_at - -
          Mouse Genome 430 2.0 Array 1429040_PM_at - -
          Mouse Genome 430 2.0 Array 1457673_PM_at - -
          Mouse Genome 430 2.0 Array 1437717_PM_x_at - -
          Mouse Genome 430 2.0 Array 1453589_a_at - -
          Mouse Genome 430 2.0 Array 1453589_PM_a_at - -
          Affymetrix Mouse Clariom S Array PSR0800014466.mm.1 TC0800003240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014466.mm.1 NR_030708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014511.mm.1 TC0800003241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014511.mm.1 NR_028428.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014473.mm.1 TC0800003240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014473.mm.1 NR_030708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014472.mm.1 TC0800003240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014472.mm.1 NR_030708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014500.mm.1 TC0800003241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014500.mm.1 NR_028428.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014514.mm.1 TC0800003241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014514.mm.1 NR_028428.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014463.mm.1 TC0800003240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014463.mm.1 NR_030708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014467.mm.1 TC0800003240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014467.mm.1 NR_030708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014482.mm.1 TC0800003240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014482.mm.1 NR_030708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014501.mm.1 TC0800003241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014501.mm.1 NR_028428.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014508.mm.1 TC0800003241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014508.mm.1 NR_028428.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014465.mm.1 TC0800003240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014465.mm.1 NR_030708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014513.mm.1 TC0800003241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014513.mm.1 NR_028428.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024775.mm.1 TC0800003241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800024775.mm.1 NR_028428.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014469.mm.1 TC0800003240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014469.mm.1 NR_030708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014476.mm.1 TC0800003240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014476.mm.1 NR_030708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014499.mm.1 TC0800003241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014499.mm.1 NR_028428.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014516.mm.1 TC0800003241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014516.mm.1 NR_028428.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014488.mm.1 TC0800003240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014488.mm.1 NR_030708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014510.mm.1 TC0800003241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014510.mm.1 NR_028428.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014515.mm.1 TC0800003241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014515.mm.1 NR_028428.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800014498.mm.1 TC0800003241.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800014498.mm.1 NR_028428.1 Refseq
          Mouse Expression Set 430 1453589_PM_a_at - -
          Mouse Expression Set 430 1453589_a_at - -
          Mouse Expression Set 430 1429040_at - -
          Mouse Expression Set 430 1452997_at - -
          Mouse Expression Set 430 1429040_PM_at - -
          Mouse Expression Set 430 1429041_at - -
          Mouse Expression Set 430 1452997_PM_at - -
          Mouse Expression Set 430 1429041_PM_at - -
          Mouse Expression Set 430 1440387_PM_at - -
          Mouse Expression Set 430 1436740_at - -
          Mouse Expression Set 430 1436740_PM_at - -
          Mouse Expression Set 430 1435470_PM_at - -
          Mouse Expression Set 430 1457673_at - -
          Mouse Expression Set 430 1440387_at - -
          Mouse Expression Set 430 1429882_PM_at - -
          Mouse Expression Set 430 1435470_at - -
          Mouse Expression Set 430 1457673_PM_at - -
          Mouse Expression Set 430 1429882_at - -
          Murine Genome U74v2 Set 103010_PM_at - -
          Murine Genome U74v2 Set 161462_r_at - -
          Murine Genome U74v2 Set 161462_PM_r_at - -
          Murine Genome U74v2 Set 103010_at - -
          Murine Genome U74v2 Set 170817_PM_f_at - -
          Murine Genome U74v2 Set 170817_f_at - -
          Murine Genome U74v2 Set 128645_PM_at - -
          Murine Genome U74v2 Set 128645_at - -
          Mouse Genome 430A 2.0 Array 1429041_at - -
          Mouse Genome 430A 2.0 Array 1453589_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1453589_a_at - -
          Mouse Genome 430A 2.0 Array 1429041_PM_at - -
          Mouse Genome 430A 2.0 Array 1429040_PM_at - -
          Mouse Genome 430A 2.0 Array 1452997_at - -
          Mouse Genome 430A 2.0 Array 1429040_at - -
          Mouse Genome 430A 2.0 Array 1452997_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1453589_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1429040_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1453589_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1429040_at - -
          Affymetrix HT Mouse Genome 430A Array 1429041_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1429041_at - -
          Affymetrix HT Mouse Genome 430A Array 1452997_at - -
          Affymetrix HT Mouse Genome 430A Array 1452997_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440387_at - -
          Affymetrix HT Mouse Genome 430B Array 1437717_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1435470_at - -
          Affymetrix HT Mouse Genome 430B Array 1437717_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1457673_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457673_at - -
          Affymetrix HT Mouse Genome 430B Array 1436740_at - -
          Affymetrix HT Mouse Genome 430B Array 1429882_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429882_at - -
          Affymetrix HT Mouse Genome 430B Array 1440387_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436740_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435470_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 451834 - -
          ABI Mouse Genome Survey Microarray 840757 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2542311 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0