Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02764817_s1
          See other GM1322 GE Assays ›
          Gene Symbol:
          Gm1322 Xirp2
          Gene Name:
          predicted gene 1322
          xin actin-binding repeat containing 2
          Gene Aliases:
          -
          2310003D02Rik, 2310008C07Rik, A530024P18Rik, AI452089, Cmya3, Gm352, Xin2
          Chromosome Location:
          Chr.2: 67173834 - 67186212 on Build GRCm38
          Chromosome Location:
          Chr.2: 67446000 - 67526620 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm02764817_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm1322
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033477.3 NP_001028649.2 2 538 80
          GenBank mRNA AK158464.1 - 2 539 80
          Gene Symbol: Xirp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA KM273012.1 - 348 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000965.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P130994 NM_001033477.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 832101 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 653410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132363 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 275222 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 103397 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633020 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 43950 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P130994 NM_001033477.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04550688 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04550700 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04550666 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04550678 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04550666 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04550700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21295809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02160290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01715120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295951 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02105575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01630964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02194870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02008972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02194874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21295809 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10472424 - -
          Affymetrix Mouse Gene 1.0 ST Array 10472423 - -
          Affymetrix Mouse Gene 1.0 ST Array 10472425 - -
          Affymetrix Mouse Gene 1.1 ST Array 10472422 - -
          Affymetrix Mouse Clariom S Array PSR0200007645.mm.1 TC0200000965.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200007645.mm.1 NM_001033477.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007645.mm.1 NM_001033477.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007644.mm.1 TC0200000965.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200007644.mm.1 NM_001033477.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007644.mm.1 NM_001033477.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007643.mm.1 TC0200000965.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200007643.mm.1 NM_001033477.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200007643.mm.1 NM_001033477.3 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161133 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161133 - -
          mouse-6 v1.1 (Illumina) GI_85702008-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_2985761 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2985761 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0