Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700002654.mm.1 |
TC0700000368.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002654.mm.1 |
NM_001045486.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002654.mm.1 |
NM_001289634.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002654.mm.1 |
NM_001289637.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002654.mm.1 |
NM_001289640.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002654.mm.1 |
NM_001289641.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002653.mm.1 |
TC0700000368.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002653.mm.1 |
NM_001289634.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002657.mm.1 |
TC0700000368.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002657.mm.1 |
NM_001045486.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002657.mm.1 |
NM_001289634.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002657.mm.1 |
NM_001289637.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002657.mm.1 |
NM_001289640.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002657.mm.1 |
NM_001289641.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002656.mm.1 |
TC0700000368.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700002656.mm.1 |
NM_001045486.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002656.mm.1 |
NM_001289634.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002656.mm.1 |
NM_001289637.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700002656.mm.1 |
NM_001289640.1 |
Refseq |