Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03015813_g1
          See other BC030499 GE Assays ›
          Gene Symbol:
          BC030499
          Gene Name:
          cDNA sequence BC030499
          Gene Aliases:
          BC037104, Sgk494
          Chromosome Location:
          Chr.11: 78290732 - 78296805 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03015813_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BC030499
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001287206.1 NP_001274135.1 8 - 9 740 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001154.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          phosphorylation
          nucleotide binding
          molecular_function
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165046 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062617 BC059191.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062617 BC030499.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062617 AK138440.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062617 BB183694.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062612 BC037104.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062612 BC048862.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07407856 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165046 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165046 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10379126 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379123 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379118 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379121 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379124 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379120 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379122 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379119 - -
          Affymetrix Mouse Gene 1.0 ST Array 10379125 - -
          Affymetrix HT MG-430 PM Array Plate 1444835_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1444656_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10379117 - -
          Mouse Genome 430 2.0 Array 1444656_PM_at - -
          Mouse Genome 430 2.0 Array 1444656_at - -
          Mouse Genome 430 2.0 Array 1444835_PM_at - -
          Mouse Genome 430 2.0 Array 1444835_at - -
          Affymetrix Mouse Clariom S Array PSR1100010678.mm.1 TC1100001154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010678.mm.1 NM_001287206.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010693.mm.1 TC1100001154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010693.mm.1 NM_001287206.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010687.mm.1 TC1100001154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010687.mm.1 NM_001287206.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010691.mm.1 TC1100001154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010691.mm.1 NM_001287206.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010686.mm.1 TC1100001154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010686.mm.1 NM_001287206.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010688.mm.1 TC1100001154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010688.mm.1 NM_001287206.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010690.mm.1 TC1100001154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010690.mm.1 NM_001287206.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010676.mm.1 TC1100001154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010676.mm.1 NM_001287206.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010689.mm.1 TC1100001154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010689.mm.1 NM_001287206.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010692.mm.1 TC1100001154.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010692.mm.1 NM_001287206.1 Refseq
          Mouse Expression Set 430 1444656_at - -
          Mouse Expression Set 430 1444656_PM_at - -
          Mouse Expression Set 430 1444835_PM_at - -
          Mouse Expression Set 430 1444835_at - -
          Affymetrix HT Mouse Genome 430B Array 1444835_at - -
          Affymetrix HT Mouse Genome 430B Array 1444656_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444656_at - -
          Affymetrix HT Mouse Genome 430B Array 1444835_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2822160 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc