Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03047322_gH
          See other PTP4A2 GE Assays ›
          Gene Symbol:
          Ptp4a2
          Gene Name:
          protein tyrosine phosphatase 4a2
          Gene Aliases:
          Prl-2
          Chromosome Location:
          Chr.4: 129820479 - 129850003 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03047322_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptp4a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164745.1 NP_001158217.1 2 - 3 795 88
          NM_008974.4 NP_033000.1 2 - 3 826 88
          GenBank mRNA AF035644.1 - 1 - 2 277 88
          AK017872.1 - 1 - 2 533 88
          AK033092.1 - 1 - 2 531 88
          AK045093.1 - 1 - 2 528 88
          AK155895.1 - 3 - 4 869 88
          BC086794.1 - 1 - 2 208 6811716 88
          BC087551.1 - 3 - 4 854 6329297 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001530.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptp4a2

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          dephosphorylation
          phosphoprotein phosphatase activity
          protein tyrosine phosphatase activity
          hydrolase activity
          phosphatase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400011873.mm.1 TC0400001530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011873.mm.1 NM_001164745.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011873.mm.1 NM_001164745.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011883.mm.1 TC0400001530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011883.mm.1 NM_001164745.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011883.mm.1 NM_001164745.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011883.mm.1 NM_008974.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011883.mm.1 NM_008974.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011877.mm.1 TC0400001530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011877.mm.1 NM_001164745.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011877.mm.1 NM_001164745.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011877.mm.1 NM_008974.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011877.mm.1 NM_008974.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011870.mm.1 TC0400001530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011870.mm.1 NM_008974.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011870.mm.1 NM_008974.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011878.mm.1 TC0400001530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011878.mm.1 NM_001164745.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011878.mm.1 NM_001164745.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011878.mm.1 NM_008974.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011878.mm.1 NM_008974.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011879.mm.1 TC0400001530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011879.mm.1 NM_001164745.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011879.mm.1 NM_001164745.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011879.mm.1 NM_008974.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011879.mm.1 NM_008974.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011880.mm.1 TC0400001530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011880.mm.1 NM_001164745.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011880.mm.1 NM_001164745.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011880.mm.1 NM_008974.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011880.mm.1 NM_008974.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011865.mm.1 TC0400001530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011869.mm.1 TC0400001530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011869.mm.1 NM_008974.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011869.mm.1 NM_008974.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011881.mm.1 TC0400001530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011881.mm.1 NM_001164745.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011881.mm.1 NM_001164745.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011881.mm.1 NM_008974.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011881.mm.1 NM_008974.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011875.mm.1 TC0400001530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011875.mm.1 NM_001164745.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011875.mm.1 NM_001164745.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011882.mm.1 TC0400001530.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400011882.mm.1 NM_001164745.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011882.mm.1 NM_001164745.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011882.mm.1 NM_008974.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400011882.mm.1 NM_008974.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 360988 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0