Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03047442_m1
          See other TNFRSF13B GE Assays ›
          Gene Symbol:
          Tnfrsf13b
          Gene Name:
          tumor necrosis factor receptor superfamily, member 13b
          Gene Aliases:
          1200009E08Rik, Taci
          Chromosome Location:
          Chr.11: 61126742 - 61149373 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03047442_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnfrsf13b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021349.1 NP_067324.1 3 - 4 522 60
          GenBank mRNA AF257673.1 - 3 - 4 522 60
          AK004668.1 - 4 - 5 599 60
          AK155178.1 - 2 - 3 206 60
          BC139392.1 - 3 - 4 579 8862414 60
          BC139393.1 - 3 - 4 579 8862415 60
          BC141867.1 - 3 - 4 522 40054144 60
          BC141868.1 - 3 - 4 522 40054146 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000774.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tnfrsf13b

          Gene Ontology Categories:

          Function(s)

          B cell homeostasis
          hematopoietic progenitor cell differentiation
          immune system process
          negative regulation of B cell proliferation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27846774 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27846773 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27846772 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981972 BC141867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981972 BC141868.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981972 AK155178.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981972 BC139392.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1981972 BC139393.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169559 BC141867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169559 BC141868.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169559 AK004668.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169559 BC139392.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169559 BC139393.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 155332 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 22735 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 942381 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86544 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981972 BC141867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981972 BC141868.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981972 AK155178.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981972 BC139392.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1981972 BC139393.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169559 BC141867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169559 BC141868.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169559 AK004668.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169559 BC139392.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169559 BC139393.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P106269 BC141867.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P106269 BC141868.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P106269 AK004668.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P106269 BC139392.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P106269 BC139393.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02340257 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02340266 - -
          Whole Mouse Genome G4122A A_51_P106269 BC141867.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P106269 BC141868.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P106269 AK004668.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P106269 BC139392.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P106269 BC139393.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129932 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100036 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27846774 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27846772 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27846773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27846767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09240216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846696 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09219807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09123465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09219802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846748 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09521726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09332855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09503315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09219805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09450601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09599172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846728 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09450599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846706 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27846709 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10376760 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376758 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376757 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376762 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376763 - -
          Affymetrix Mouse Gene 1.0 ST Array 10376759 - -
          Affymetrix HT MG-430 PM Array Plate 1423182_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10376755 - -
          Mouse Genome 430 2.0 Array 1423182_PM_at - -
          Mouse Genome 430 2.0 Array 1423182_at - -
          Affymetrix Mouse Clariom S Array PSR1100006204.mm.1 TC1100000774.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100006203.mm.1 TC1100000774.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100006203.mm.1 NM_021349.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100006203.mm.1 NM_021349.1 Refseq
          Affymetrix HT Mouse Genome 430A Array 1423182_at - -
          Affymetrix HT Mouse Genome 430A Array 1423182_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 642510 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102640397 - -
          Illumina mouse-6 v1.0 expression beadchip 1990373 - -
          Illumina mouse-6 v1.0 expression beadchip 102640397 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001990373 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2480814 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2480814 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2422416 - -
          mouse-6 v1.1 (Illumina) scl41469.8_427-S - -
          mouse-6 v1.1 (Illumina) scl0057916.2_218-S - -
          Illumina Sentrix mouse-6.v1 bead array 102640397 - -
          Illumina Sentrix mouse-6.v1 bead array 1990373 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0057916.2_218-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1990373 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102640397 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2480814 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2480814 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2422416 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0