Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03455705_m1
          See other NESPAS GE Assays ›
          Gene Symbol:
          Nespas
          Gene Name:
          neuroendocrine secretory protein antisense
          Gene Aliases:
          Gnas-as, Nespos
          Chromosome Location:
          Chr.2: 174281237 - 174295436 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03455705_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nespas
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002846.2
          4 - 5 981 62
          GenBank mRNA AF319945.1 - LIT1669
          Nespas
          4 - 5 952 62
          AK153813.1 -
          4 - 5 982 62

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 298 Mir298 Mirn298,
          mir-298,
          mmu-mir-298
          -14.6kb Sense Chr. 2 - 174267504 - 174267585 on NCBI Build GRCm38
          Downstream GNAS (guanine nucleotide binding protein,
          alpha stimulating) complex locus
          Gnas 5530400H20Rik,
          A930027G11Rik,
          C130027O20Rik,
          GPSA,
          GSP,
          Galphas,
          Gnas1,
          Gnasxl,
          Gsa,
          Nesp,
          Nespl,
          Oed-Sml,
          Oedsml,
          P1,
          P2,
          P3,
          PHP1A,
          PHP1B,
          POH
          20.0 Anti-sense Chr. 2 - 174283444 - 174346744 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005308.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s)

          tissue homeostasis
          DNA methylation
          post-embryonic development
          multicellular organism growth
          skin development
          genetic imprinting

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P113708 AF319945.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P113708 NR_002846.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P113708 AK153813.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973595 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409634 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 440094 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 172979 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586313 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669069 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 234241 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297608 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 258269 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118256 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P113708 AF319945.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P113708 NR_002846.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P113708 AK153813.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04796605 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113708 AF319945.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113708 NR_002846.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113708 AK153813.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1427788_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56802_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56802_at - -
          Affymetrix Mouse Clariom S Array PSR0200045288.mm.1 TC0200005308.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200045288.mm.1 NR_002846.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045288.mm.1 NR_002846.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045287.mm.1 TC0200005308.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200045287.mm.1 NR_002846.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045287.mm.1 NR_002846.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045286.mm.1 TC0200005308.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200045286.mm.1 NR_002846.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200045286.mm.1 NR_002846.2 Refseq
          Murine Genome U74v2 Set 160097_at - -
          Murine Genome U74v2 Set 160097_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427788_at - -
          Affymetrix HT Mouse Genome 430A Array 1427788_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 493549 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2493616 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0