Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03455851_s1
          See other MIAT GE Assays ›
          Gene Symbol:
          Miat
          Gene Name:
          myocardial infarction associated transcript (non-protein coding)
          Gene Aliases:
          3632434I06, A230057G18Rik, Rncr2, gomafu
          Chromosome Location:
          Chr.5: 112213228 - 112228948 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03455851_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Miat
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003718.2
          7 8614 80
          NR_033657.1
          7 8658 80
          GenBank mRNA AB300594.1 -
          7 8601 80
          AK028326.1 - LIT3370
          RNCR2
          1 2956 80
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY642170.1 - 1 208 Chr. 5 - 112213230 - 112213536 [-] on Build GRCm38 80
          GenBankmRNA BE949942.1 - 1 Chr. 5 - 112213226 - 112213628 [+] on Build GRCm38 80
          GenBankmRNA CJ243523.1 - 1 236 Chr. 5 - 112213229 - 112213564 [-] on Build GRCm38 80
          GenBankmRNA GH454614.1 - Chr. - - [-] on Build 80
          GenBankmRNA AW050310.1 - 1 Chr. 5 - 112213226 - 112213543 [+] on Build GRCm38 80
          GenBankmRNA CJ242510.1 - 1 186 Chr. 5 - 112213230 - 112213515 [-] on Build GRCm38 80
          GenBankmRNA BB750006.1 - 1 338 Chr. 5 - 112213230 - 112213667 [-] on Build GRCm38 80
          GenBankmRNA BU614259.1 - 1 161 Chr. 5 - 112213226 - 112213484 [-] on Build GRCm38 80
          GenBankmRNA BX523646.1 - 1 257 Chr. 5 - 112213226 - 112213585 [-] on Build GRCm38 80
          GenBankmRNA BB466195.1 - 1 179 Chr. 5 - 112213228 - 112213508 [-] on Build GRCm38 80
          GenBankmRNA CJ242470.1 - 1 183 Chr. 5 - 112213229 - 112213512 [-] on Build GRCm38 80
          GenBankmRNA CJ243319.1 - 1 203 Chr. 5 - 112213230 - 112213532 [-] on Build GRCm38 80
          GenBankmRNA CO043947.1 - 1 Chr. 5 - 112213226 - 112213504 [+] on Build GRCm38 80
          GenBankmRNA CJ243480.1 - 1 237 Chr. 5 - 112213230 - 112213565 [-] on Build GRCm38 80
          GenBankmRNA BB672243.1 - 1 280 Chr. 5 - 112213232 - 112213602 [-] on Build GRCm38 80
          GenBankmRNA CJ242530.1 - 1 182 Chr. 5 - 112213229 - 112213511 [-] on Build GRCm38 80
          GenBankmRNA BX525971.1 - 1 335 Chr. 5 - 112213226 - 112213664 [-] on Build GRCm38 80
          GenBankmRNA W59359.1 - 280 Chr. - - [-] on Build 80
          GenBankmRNA AV221928.2 - 1 571 Chr. 5 - 112213230 - 112213875 [-] on Build GRCm38 80
          GenBankmRNA CJ243570.1 - 1 307 Chr. 5 - 112213230 - 112213635 [-] on Build GRCm38 80
          GenBankmRNA CJ244888.1 - 1 356 Chr. 5 - 112213230 - 112213673 [-] on Build GRCm38 80
          GenBankmRNA AB294517.1 - 1 Chr. 5 - 112213226 - 112213579 [+] on Build GRCm38 80
          GenBankmRNA BE951950.1 - 1 Chr. 5 - 112213226 - 112213503 [+] on Build GRCm38 80
          GenBankmRNA W62221.1 - 277 Chr. - - [-] on Build 80
          GenBankmRNA CJ242762.1 - 1 205 Chr. 5 - 112213229 - 112213534 [-] on Build GRCm38 80
          GenBankmRNA AA270931.1 - 1 254 Chr. 5 - 112213241 - 112213585 [-] on Build GRCm38 80
          GenBankmRNA CJ242581.1 - 1 206 Chr. 5 - 112213230 - 112213534 [-] on Build GRCm38 80
          GenBankmRNA CJ242518.1 - 1 324 Chr. 5 - 112213230 - 112213652 [-] on Build GRCm38 80
          GenBankmRNA CJ242517.1 - 1 209 Chr. 5 - 112213230 - 112213538 [-] on Build GRCm38 80
          GenBankmRNA CR517013.1 - 1 Chr. 5 - 112213226 - 112213568 [+] on Build GRCm38 80
          GenBankmRNA BB100360.1 - 1 170 Chr. 5 - 112213232 - 112213499 [-] on Build GRCm38 80
          GenBankmRNA BB320450.1 - 1 219 Chr. 5 - 112213230 - 112213513 [-] on Build GRCm38 80

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 1700028D13 gene 1700028D13Rik -3.2kb Anti-sense Chr. 5 - 112206763 - 112210002 on NCBI Build GRCm38
          Downstream crystallin,
          beta A4
          Cryba4 30.2kb Sense Chr. 5 - 112246492 - 112263555 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003012.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          glial cell differentiation
          stem cell population maintenance
          amacrine cell differentiation
          retinal rod cell development
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2128860 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806023 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 661521 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 21327 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855232 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 794292 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 742705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 27800 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 52231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04807417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04807423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23990687 - -
          Array Name Probe IDs Targets Target Type
          Mouse Genome 430 2.0 Array 1447262_at - -
          Mouse Genome 430 2.0 Array 1447262_PM_at - -
          Mouse Genome 430 2.0 Array 1458903_PM_at - -
          Mouse Genome 430 2.0 Array 1455325_at - -
          Mouse Genome 430 2.0 Array 1455325_PM_at - -
          Mouse Genome 430 2.0 Array 1458903_at - -
          Affymetrix Mouse Clariom S Array PSR0500026694.mm.1 TC0500003012.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026694.mm.1 NR_003718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026694.mm.1 NR_003718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026694.mm.1 NR_033657.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026694.mm.1 NR_033657.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026697.mm.1 TC0500003012.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026697.mm.1 NR_003718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026697.mm.1 NR_003718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026697.mm.1 NR_033657.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026697.mm.1 NR_033657.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026695.mm.1 TC0500003012.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026695.mm.1 NR_003718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026695.mm.1 NR_003718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026695.mm.1 NR_033657.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026695.mm.1 NR_033657.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026693.mm.1 TC0500003012.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026693.mm.1 NR_003718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026693.mm.1 NR_003718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026693.mm.1 NR_033657.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026693.mm.1 NR_033657.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026696.mm.1 TC0500003012.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026696.mm.1 NR_003718.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026696.mm.1 NR_003718.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026696.mm.1 NR_033657.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026696.mm.1 NR_033657.1 Refseq
          Mouse Expression Set 430 1455325_PM_at - -
          Mouse Expression Set 430 1455325_at - -
          Mouse Expression Set 430 1458903_at - -
          Mouse Expression Set 430 1458903_PM_at - -
          Murine Genome U74v2 Set 167615_PM_s_at - -
          Murine Genome U74v2 Set 139503_PM_at - -
          Murine Genome U74v2 Set 167615_s_at - -
          Murine Genome U74v2 Set 139503_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 471664 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c8579b687-54cxl:80/100.66.135.11:80.
          git-commit: 8e87205ac1279514f9c7a0d34bcd4a43061862b7
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0