Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03456006_m1
          See other EMX2OS GE Assays ›
          Gene Symbol:
          Emx2os
          Gene Name:
          Emx2 opposite strand/antisense transcript (non-protein coding)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 59425104 - 59458635 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03456006_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Emx2os
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002863.2
          3 - 4 863 118
          GenBank mRNA AK033011.1 -
          3 - 4 512 118
          AY117414.1 - LIT1111
          EMX2OS
          3 - 4 862 118

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream PDZ domain containing 8 Pdzd8 A630041P07Rik,
          AV329468,
          AW228944,
          Pdzk8
          -79.3kb Sense Chr. 19 - 59296084 - 59345780 on NCBI Build GRCm38
          Downstream empty spiracles homeobox 2 Emx2 Pdo 28.0kb Anti-sense Chr. 19 - 59458116 - 59465357 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001734.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186460 NR_002863.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186460 AK033011.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2186460 AY117414.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994062 NR_002863.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994062 AY117414.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755673 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587616 AB041045.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587616 AK030737.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587616 BC017144.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587616 BC060654.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587616 NM_177699.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 778509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 538626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 387158 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 15353 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746549 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 153020 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 37320 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 381146 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 147238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 520638 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 918473 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427322 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 171815 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 625256 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 631779 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790066 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 100769 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 19862 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 10579 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 7328 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178115 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09217481 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09217493 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09217461 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178115 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03629899 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104983 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09217493 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178115 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09217461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14200979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219419 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14336100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219399 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14436185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14503079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219425 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14477708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14282696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14198624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14468266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14351711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219405 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14328154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14282694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14282015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219298 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14400664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14328155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14240384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14420154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14373151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14468265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14436186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219363 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219442 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14503072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219413 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14436181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219416 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32219272 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1456559_PM_at - -
          Mouse Genome 430 2.0 Array 1456559_at - -
          Mouse Genome 430 2.0 Array 1456559_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 329078_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 329078_at - -
          Affymetrix Mouse Clariom S Array PSR1900015356.mm.1 TC1900001734.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900015356.mm.1 NR_002863.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900015356.mm.1 NR_002863.2 Refseq
          Mouse Expression Set 430 1456559_at - -
          Mouse Expression Set 430 1456559_PM_at - -
          Murine Genome U74v2 Set 167831_PM_at - -
          Murine Genome U74v2 Set 167831_at - -
          Affymetrix HT Mouse Genome 430B Array 1456559_at - -
          Affymetrix HT Mouse Genome 430B Array 1456559_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 429490 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2429356 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0